Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12949552-12949777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM234B
Entrez Gene ID
57613
Gene Name
family with sequence similarity 234 member B
Gene Aliases
KIAA1467
UniGene
Hs.132660
Gene Chromosome Location
Chr.12: 12927680-13083449 on Build GRCh38
Transcripts
XM_017019706.1 NM_018654.1 XM_005253421.3 XM_017019582.1 XM_017019583.1

Gene Symbol
GPRC5D
Entrez Gene ID
55507
Gene Name
G protein-coupled receptor class C group 5 member D
Gene Aliases
-
UniGene
Hs.644599
Gene Chromosome Location
Chr.12: 12940518-12955908 on Build GRCh38
Transcripts
XM_017019706.1 NM_018654.1 XM_005253421.3 XM_017019582.1 XM_017019583.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4455154 c.683A>G p.N228S Chr.12: 12949702-12949702 on Build GRCh38
6674608 c.723C>T p.V241V Chr.12: 12949662-12949662 on Build GRCh38
4414243 c.761T>A p.L254Q Chr.12: 12949624-12949624 on Build GRCh38
1208791 c.755T>G p.V252G Chr.12: 12949630-12949630 on Build GRCh38
4172076 c.754G>C p.V252L Chr.12: 12949631-12949631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750967244 Chr.12: 12949638-12949638 on Build GRCh38
rs764779499 Chr.12: 12949619-12949619 on Build GRCh38
rs754518641 Chr.12: 12949613-12949613 on Build GRCh38
rs764549003 Chr.12: 12949589-12949589 on Build GRCh38
rs376715622 Chr.12: 12949585-12949585 on Build GRCh38
rs748341471 Chr.12: 12949753-12949753 on Build GRCh38
rs373330973 Chr.12: 12949666-12949666 on Build GRCh38
rs768737138 Chr.12: 12949662-12949662 on Build GRCh38
rs369458282 Chr.12: 12949713-12949713 on Build GRCh38
rs201518268 Chr.12: 12949701-12949701 on Build GRCh38
rs777649046 Chr.12: 12949628-12949628 on Build GRCh38
rs746007168 Chr.12: 12949645-12949645 on Build GRCh38
rs752208918 Chr.12: 12949681-12949681 on Build GRCh38
rs138146014 Chr.12: 12949587-12949587 on Build GRCh38
rs775483843 Chr.12: 12949659-12949659 on Build GRCh38
rs752399230 Chr.12: 12949620-12949620 on Build GRCh38
rs751439160 Chr.12: 12949699-12949699 on Build GRCh38
rs202240438 Chr.12: 12949688-12949688 on Build GRCh38
rs745916681 Chr.12: 12949578-12949578 on Build GRCh38
rs746762439 Chr.12: 12949637-12949637 on Build GRCh38
rs201988511 Chr.12: 12949670-12949670 on Build GRCh38
rs763626866 Chr.12: 12949698-12949698 on Build GRCh38
rs773866153 Chr.12: 12949672-12949672 on Build GRCh38
rs753384536 Chr.12: 12949600-12949600 on Build GRCh38
rs758087229 Chr.12: 12949623-12949623 on Build GRCh38
rs762431769 Chr.12: 12949608-12949608 on Build GRCh38
rs370153735 Chr.12: 12949661-12949661 on Build GRCh38
rs142627612 Chr.12: 12949611-12949611 on Build GRCh38
rs761194902 Chr.12: 12949676-12949676 on Build GRCh38
rs767880543 Chr.12: 12949610-12949610 on Build GRCh38
rs143364840 Chr.12: 12949665-12949665 on Build GRCh38
rs770069414 Chr.12: 12949579-12949579 on Build GRCh38
rs201124445 Chr.12: 12949723-12949723 on Build GRCh38
rs547334306 Chr.12: 12949700-12949700 on Build GRCh38
rs764873589 Chr.12: 12949677-12949677 on Build GRCh38
rs377356062 Chr.12: 12949706-12949706 on Build GRCh38
rs149059133 Chr.12: 12949642-12949642 on Build GRCh38
rs369640440 Chr.12: 12949581-12949581 on Build GRCh38
rs147555265 Chr.12: 12949749-12949749 on Build GRCh38
rs368865547 Chr.12: 12949687-12949687 on Build GRCh38
rs139583176 Chr.12: 12949639-12949639 on Build GRCh38
rs146025174 Chr.12: 12949635-12949635 on Build GRCh38
rs4630382 Chr.12: 12949671-12949671 on Build GRCh38
rs867789644 Chr.12: 12949715-12949715 on Build GRCh38
rs774529747 Chr.12: 12949590-12949590 on Build GRCh38
rs145033785 Chr.12: 12949720-12949720 on Build GRCh38
rs373129195 Chr.12: 12949717-12949717 on Build GRCh38
rs144517754 Chr.12: 12949656-12949656 on Build GRCh38

View Full Product Details