Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74881839-74882066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLST
Entrez Gene ID
1743
Gene Name
dihydrolipoamide S-succinyltransferase
Gene Aliases
DLTS
UniGene
Hs.525459
Gene Chromosome Location
Chr.14: 74881891-74903747 on Build GRCh38
Transcripts
NM_001244883.1 NM_001933.4 NR_033814.1 NR_045209.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6554528 c.4C>T p.L2L Chr.14: 74881957-74881957 on Build GRCh38
6437277 c.8C>T p.S3F Chr.14: 74881961-74881961 on Build GRCh38
7233231 c.12A>G p.R4R Chr.14: 74881965-74881965 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762731731 Chr.14: 74881904-74881904 on Build GRCh38
rs1021384202 Chr.14: 74881963-74881963 on Build GRCh38
rs374911682 Chr.14: 74881948-74881948 on Build GRCh38
rs28364622 Chr.14: 74881943-74881943 on Build GRCh38
rs910565762 Chr.14: 74881979-74881979 on Build GRCh38
rs924714941 Chr.14: 74881879-74881879 on Build GRCh38
rs376399767 Chr.14: 74881938-74881938 on Build GRCh38
rs757382191 Chr.14: 74881909-74881909 on Build GRCh38
rs914408056 Chr.14: 74881866-74881866 on Build GRCh38
rs544892049 Chr.14: 74881969-74881969 on Build GRCh38
rs977545196 Chr.14: 74881875-74881875 on Build GRCh38
rs991373140 Chr.14: 74881951-74881951 on Build GRCh38
rs373129809 Chr.14: 74881965-74881965 on Build GRCh38
rs151007114 Chr.14: 74881993-74881993 on Build GRCh38
rs369001394 Chr.14: 74881932-74881932 on Build GRCh38
rs771965585 Chr.14: 74882035-74882035 on Build GRCh38
rs559601136 Chr.14: 74881961-74881961 on Build GRCh38
rs758674697 Chr.14: 74881983-74881983 on Build GRCh38
rs770437870 Chr.14: 74881930-74881930 on Build GRCh38
rs972782753 Chr.14: 74881870-74881870 on Build GRCh38
rs774943636 Chr.14: 74881964-74881964 on Build GRCh38
rs767241815 Chr.14: 74882049-74882049 on Build GRCh38
rs958647658 Chr.14: 74881863-74881863 on Build GRCh38
rs3207434 Chr.14: 74882004-74882004 on Build GRCh38
rs368528502 Chr.14: 74881914-74881914 on Build GRCh38
rs779183058 Chr.14: 74881927-74881927 on Build GRCh38
rs139599696 Chr.14: 74882001-74882001 on Build GRCh38
rs768282930 Chr.14: 74882026-74882026 on Build GRCh38
rs751777011 Chr.14: 74881908-74881908 on Build GRCh38
rs766735834 Chr.14: 74881972-74881972 on Build GRCh38
rs749871935 Chr.14: 74881921-74881921 on Build GRCh38
rs778158079 Chr.14: 74881990-74881990 on Build GRCh38
rs201221488 Chr.14: 74881947-74881947 on Build GRCh38
rs755094737 Chr.14: 74881980-74881980 on Build GRCh38
rs761636416 Chr.14: 74881923-74881923 on Build GRCh38
rs774155136 Chr.14: 74881911-74881912 on Build GRCh38
rs374676630 Chr.14: 74881928-74881928 on Build GRCh38
rs377341812 Chr.14: 74881967-74881967 on Build GRCh38
rs769359439 Chr.14: 74881935-74881935 on Build GRCh38
rs748168146 Chr.14: 74882033-74882033 on Build GRCh38
rs1005766540 Chr.14: 74881936-74881936 on Build GRCh38
rs753807796 Chr.14: 74881978-74881978 on Build GRCh38
rs987451544 Chr.14: 74881975-74881975 on Build GRCh38
rs776307115 Chr.14: 74881900-74881900 on Build GRCh38
rs765595175 Chr.14: 74881981-74881981 on Build GRCh38
rs750745041 Chr.14: 74881944-74881944 on Build GRCh38
rs745576936 Chr.14: 74881960-74881960 on Build GRCh38
rs879032344 Chr.14: 74881994-74881994 on Build GRCh38
rs372592399 Chr.14: 74881941-74881941 on Build GRCh38
rs955341723 Chr.14: 74881883-74881883 on Build GRCh38
rs750476967 Chr.14: 74881912-74881912 on Build GRCh38
rs759091014 Chr.14: 74882046-74882046 on Build GRCh38
rs776278156 Chr.14: 74881931-74881931 on Build GRCh38
rs1035597723 Chr.14: 74881858-74881858 on Build GRCh38
rs755397855 Chr.14: 74881949-74881949 on Build GRCh38
rs763845929 Chr.14: 74881906-74881906 on Build GRCh38
rs369774111 Chr.14: 74882032-74882032 on Build GRCh38
rs760566745 Chr.14: 74882045-74882045 on Build GRCh38
rs765337358 Chr.14: 74881933-74881933 on Build GRCh38
rs753313643 Chr.14: 74881982-74881982 on Build GRCh38
rs564767822 Chr.14: 74882042-74882042 on Build GRCh38
rs528421129 Chr.14: 74881966-74881966 on Build GRCh38
rs540679704 Chr.14: 74881910-74881910 on Build GRCh38
rs538914771 Chr.14: 74881873-74881873 on Build GRCh38
rs920871868 Chr.14: 74881985-74881985 on Build GRCh38
rs779695589 Chr.14: 74881995-74881995 on Build GRCh38
rs200502397 Chr.14: 74881950-74881950 on Build GRCh38
rs991279742 Chr.14: 74881864-74881864 on Build GRCh38
rs771215689 Chr.14: 74882047-74882047 on Build GRCh38
rs780704488 Chr.14: 74881917-74881917 on Build GRCh38

View Full Product Details