Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167063515-167063774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPA33
Entrez Gene ID
10223
Gene Name
glycoprotein A33
Gene Aliases
A33
UniGene
Hs.651244
Gene Chromosome Location
Chr.1: 167052836-167090631 on Build GRCh38
Transcripts
NM_005814.2 XM_017000005.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
340152 c.562G>T p.A188S Chr.1: 167063591-167063591 on Build GRCh38
6122379 c.463G>T p.G155W Chr.1: 167063690-167063690 on Build GRCh38
6672950 c.444C>T p.I148I Chr.1: 167063709-167063709 on Build GRCh38
3863408 c.496G>A p.E166K Chr.1: 167063657-167063657 on Build GRCh38
6672952 c.500G>A p.G167D Chr.1: 167063653-167063653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944133476 Chr.1: 167063720-167063720 on Build GRCh38
rs751598020 Chr.1: 167063682-167063682 on Build GRCh38
rs202193771 Chr.1: 167063688-167063688 on Build GRCh38
rs781019912 Chr.1: 167063619-167063619 on Build GRCh38
rs754860161 Chr.1: 167063616-167063616 on Build GRCh38
rs373807305 Chr.1: 167063539-167063539 on Build GRCh38
rs577237818 Chr.1: 167063712-167063712 on Build GRCh38
rs767246696 Chr.1: 167063695-167063695 on Build GRCh38
rs918352769 Chr.1: 167063561-167063561 on Build GRCh38
rs113734455 Chr.1: 167063618-167063618 on Build GRCh38
rs149079321 Chr.1: 167063671-167063671 on Build GRCh38
rs774532389 Chr.1: 167063741-167063741 on Build GRCh38
rs748736699 Chr.1: 167063641-167063641 on Build GRCh38
rs760028529 Chr.1: 167063572-167063573 on Build GRCh38
rs746744733 Chr.1: 167063547-167063547 on Build GRCh38
rs992363391 Chr.1: 167063681-167063681 on Build GRCh38
rs759396228 Chr.1: 167063745-167063745 on Build GRCh38
rs763245601 Chr.1: 167063733-167063733 on Build GRCh38
rs891244148 Chr.1: 167063714-167063714 on Build GRCh38
rs768947624 Chr.1: 167063657-167063657 on Build GRCh38
rs756795354 Chr.1: 167063708-167063708 on Build GRCh38
rs576714423 Chr.1: 167063665-167063665 on Build GRCh38
rs200961071 Chr.1: 167063611-167063611 on Build GRCh38
rs758427123 Chr.1: 167063597-167063597 on Build GRCh38
rs767868865 Chr.1: 167063641-167063641 on Build GRCh38
rs777150420 Chr.1: 167063628-167063628 on Build GRCh38
rs148089044 Chr.1: 167063713-167063713 on Build GRCh38
rs747479502 Chr.1: 167063647-167063647 on Build GRCh38
rs776755193 Chr.1: 167063593-167063593 on Build GRCh38
rs146018358 Chr.1: 167063675-167063675 on Build GRCh38
rs750453067 Chr.1: 167063596-167063596 on Build GRCh38
rs780601775 Chr.1: 167063644-167063644 on Build GRCh38
rs769236043 Chr.1: 167063554-167063554 on Build GRCh38
rs772729172 Chr.1: 167063557-167063557 on Build GRCh38
rs753482303 Chr.1: 167063706-167063706 on Build GRCh38
rs376202132 Chr.1: 167063589-167063589 on Build GRCh38
rs367726896 Chr.1: 167063549-167063549 on Build GRCh38
rs141668937 Chr.1: 167063696-167063696 on Build GRCh38
rs747777864 Chr.1: 167063621-167063621 on Build GRCh38
rs761083043 Chr.1: 167063575-167063575 on Build GRCh38
rs777536644 Chr.1: 167063703-167063703 on Build GRCh38
rs762050223 Chr.1: 167063594-167063594 on Build GRCh38
rs755733404 Chr.1: 167063701-167063701 on Build GRCh38
rs539164220 Chr.1: 167063548-167063548 on Build GRCh38
rs767429910 Chr.1: 167063748-167063748 on Build GRCh38
rs2228399 Chr.1: 167063658-167063658 on Build GRCh38
rs375084264 Chr.1: 167063567-167063567 on Build GRCh38
rs12742013 Chr.1: 167063559-167063559 on Build GRCh38
rs772500496 Chr.1: 167063560-167063560 on Build GRCh38
rs140361439 Chr.1: 167063664-167063664 on Build GRCh38
rs771158351 Chr.1: 167063739-167063739 on Build GRCh38
rs766535525 Chr.1: 167063676-167063676 on Build GRCh38
rs377665923 Chr.1: 167063571-167063571 on Build GRCh38
rs778368595 Chr.1: 167063709-167063709 on Build GRCh38
rs766484031 Chr.1: 167063608-167063608 on Build GRCh38
rs770410900 Chr.1: 167063643-167063643 on Build GRCh38
rs188793256 Chr.1: 167063723-167063723 on Build GRCh38
rs369125005 Chr.1: 167063728-167063728 on Build GRCh38
rs775843538 Chr.1: 167063565-167063565 on Build GRCh38
rs2228399 Chr.1: 167063658-167063658 on Build GRCh38
rs575939444 Chr.1: 167063715-167063715 on Build GRCh38

View Full Product Details