Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63346813-63347084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA4
Entrez Gene ID
1137
Gene Name
cholinergic receptor nicotinic alpha 4 subunit
Gene Aliases
BFNC, EBN, EBN1, NACHR, NACHRA4, NACRA4
UniGene
Hs.10734
Gene Chromosome Location
Chr.20: 63343310-63375392 on Build GRCh38
Transcripts
NM_000744.6 NM_001256573.1 NR_046317.1 XM_011528524.1 XM_017027624.1 XM_017027625.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7130056 c.1806C>T p.F602F Chr.20: 63346816-63346816 on Build GRCh38
1661140 c.1759G>A p.V587M Chr.20: 63346863-63346863 on Build GRCh38
2931853 c.1765G>A p.E589K Chr.20: 63346857-63346857 on Build GRCh38
6300450 c.1759-4G>A p.? Chr.20: 63346867-63346867 on Build GRCh38
356954 c.1775A>C p.K592T Chr.20: 63346847-63346847 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370147172 Chr.20: 63346882-63346882 on Build GRCh38
rs897203483 Chr.20: 63346932-63346932 on Build GRCh38
rs983786313 Chr.20: 63347059-63347059 on Build GRCh38
rs370887602 Chr.20: 63346842-63346842 on Build GRCh38
rs749939383 Chr.20: 63346857-63346857 on Build GRCh38
rs113592412 Chr.20: 63346912-63346912 on Build GRCh38
rs199842495 Chr.20: 63346894-63346894 on Build GRCh38
rs368125570 Chr.20: 63346881-63346881 on Build GRCh38
rs201735615 Chr.20: 63346880-63346880 on Build GRCh38
rs45490595 Chr.20: 63346937-63346937 on Build GRCh38
rs45440192 Chr.20: 63346877-63346877 on Build GRCh38
rs914220324 Chr.20: 63347046-63347046 on Build GRCh38
rs561318608 Chr.20: 63346891-63346891 on Build GRCh38
rs751163361 Chr.20: 63346878-63346878 on Build GRCh38
rs993730782 Chr.20: 63347023-63347023 on Build GRCh38
rs879125634 Chr.20: 63346876-63346876 on Build GRCh38
rs1008121764 Chr.20: 63346999-63346999 on Build GRCh38
rs953056734 Chr.20: 63346936-63346936 on Build GRCh38
rs569285028 Chr.20: 63347040-63347040 on Build GRCh38
rs958507188 Chr.20: 63347062-63347062 on Build GRCh38
rs200251092 Chr.20: 63346906-63346906 on Build GRCh38
rs1025225297 Chr.20: 63347034-63347034 on Build GRCh38
rs56328826 Chr.20: 63346893-63346893 on Build GRCh38
rs369313655 Chr.20: 63346933-63346933 on Build GRCh38
rs200965304 Chr.20: 63346898-63346898 on Build GRCh38
rs374354698 Chr.20: 63346867-63346867 on Build GRCh38
rs187614771 Chr.20: 63346985-63346985 on Build GRCh38
rs1041070823 Chr.20: 63346916-63346916 on Build GRCh38
rs756800196 Chr.20: 63347039-63347039 on Build GRCh38
rs765835508 Chr.20: 63346905-63346905 on Build GRCh38
rs547896102 Chr.20: 63347018-63347018 on Build GRCh38
rs200698041 Chr.20: 63346836-63346836 on Build GRCh38
rs538150310 Chr.20: 63347059-63347059 on Build GRCh38
rs182964504 Chr.20: 63346974-63346974 on Build GRCh38
rs762379814 Chr.20: 63346911-63346911 on Build GRCh38
rs988457885 Chr.20: 63347041-63347041 on Build GRCh38
rs1027265354 Chr.20: 63346935-63346935 on Build GRCh38
rs747216106 Chr.20: 63346899-63346899 on Build GRCh38
rs768939689 Chr.20: 63346902-63346902 on Build GRCh38
rs536332563 Chr.20: 63347002-63347002 on Build GRCh38
rs201347598 Chr.20: 63346843-63346843 on Build GRCh38
rs928360708 Chr.20: 63347025-63347025 on Build GRCh38
rs200032652 Chr.20: 63346862-63346862 on Build GRCh38
rs1024236176 Chr.20: 63346939-63346939 on Build GRCh38
rs267606047 Chr.20: 63346849-63346849 on Build GRCh38
rs373231502 Chr.20: 63347061-63347061 on Build GRCh38
rs777310362 Chr.20: 63346839-63346839 on Build GRCh38
rs1000245569 Chr.20: 63346972-63346972 on Build GRCh38
rs934274812 Chr.20: 63346887-63346887 on Build GRCh38
rs889244653 Chr.20: 63346992-63346992 on Build GRCh38
rs763684538 Chr.20: 63346903-63346903 on Build GRCh38

View Full Product Details