Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19055752-19055981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC6
Entrez Gene ID
93436
Gene Name
armadillo repeat containing 6
Gene Aliases
R30923_1
UniGene
Hs.77876
Gene Chromosome Location
Chr.19: 19033578-19058178 on Build GRCh38
Transcripts
NM_001199196.1 NM_033415.3 XM_005260157.3 XM_017027484.1 XM_017027485.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3530953 c.1192G>A p.A398T Chr.19: 19055902-19055902 on Build GRCh38
3100775 c.1236C>T p.G412G Chr.19: 19055871-19055871 on Build GRCh38
6343510 c.1081-10G>T p.? Chr.19: 19055781-19055781 on Build GRCh38
5959096 c.1272delC p.P425fs*13 Chr.19: 19055907-19055907 on Build GRCh38
6335938 c.1131C>T p.P377P Chr.19: 19055841-19055841 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772707062 Chr.19: 19055888-19055888 on Build GRCh38
rs376475842 Chr.19: 19055775-19055775 on Build GRCh38
rs748357975 Chr.19: 19055844-19055844 on Build GRCh38
rs778200629 Chr.19: 19055919-19055919 on Build GRCh38
rs28729373 Chr.19: 19055781-19055781 on Build GRCh38
rs765886715 Chr.19: 19055906-19055906 on Build GRCh38
rs148509679 Chr.19: 19055852-19055852 on Build GRCh38
rs749200716 Chr.19: 19055929-19055929 on Build GRCh38
rs763812983 Chr.19: 19055808-19055808 on Build GRCh38
rs776804766 Chr.19: 19055811-19055811 on Build GRCh38
rs762867708 Chr.19: 19055806-19055806 on Build GRCh38
rs753049335 Chr.19: 19055916-19055916 on Build GRCh38
rs151031705 Chr.19: 19055910-19055910 on Build GRCh38
rs775443484 Chr.19: 19055853-19055853 on Build GRCh38
rs761506360 Chr.19: 19055874-19055874 on Build GRCh38
rs368147693 Chr.19: 19055901-19055901 on Build GRCh38
rs779150296 Chr.19: 19055842-19055842 on Build GRCh38
rs761404630 Chr.19: 19055810-19055810 on Build GRCh38
rs369392289 Chr.19: 19055923-19055923 on Build GRCh38
rs756585012 Chr.19: 19055832-19055832 on Build GRCh38
rs754303232 Chr.19: 19055909-19055909 on Build GRCh38
rs142757655 Chr.19: 19055871-19055871 on Build GRCh38
rs758659488 Chr.19: 19055917-19055917 on Build GRCh38
rs775424006 Chr.19: 19055800-19055800 on Build GRCh38
rs749190903 Chr.19: 19055859-19055859 on Build GRCh38
rs754137972 Chr.19: 19055838-19055838 on Build GRCh38
rs757342971 Chr.19: 19055928-19055928 on Build GRCh38
rs781508584 Chr.19: 19055930-19055931 on Build GRCh38
rs777974631 Chr.19: 19055846-19055846 on Build GRCh38
rs768461003 Chr.19: 19055860-19055860 on Build GRCh38
rs200832309 Chr.19: 19055851-19055851 on Build GRCh38
rs769898413 Chr.19: 19055793-19055793 on Build GRCh38
rs751161347 Chr.19: 19055809-19055809 on Build GRCh38
rs142084096 Chr.19: 19055833-19055833 on Build GRCh38
rs766965311 Chr.19: 19055823-19055823 on Build GRCh38
rs755311283 Chr.19: 19055841-19055841 on Build GRCh38
rs760234596 Chr.19: 19055896-19055896 on Build GRCh38
rs771698197 Chr.19: 19055950-19055950 on Build GRCh38
rs767199863 Chr.19: 19055886-19055886 on Build GRCh38
rs746277603 Chr.19: 19055931-19055948 on Build GRCh38
rs547962054 Chr.19: 19055944-19055944 on Build GRCh38
rs749985741 Chr.19: 19055831-19055831 on Build GRCh38
rs201382714 Chr.19: 19055949-19055949 on Build GRCh38
rs768526435 Chr.19: 19055936-19055936 on Build GRCh38
rs765584331 Chr.19: 19055913-19055913 on Build GRCh38
rs759495206 Chr.19: 19055776-19055776 on Build GRCh38
rs772792845 Chr.19: 19055959-19055959 on Build GRCh38

View Full Product Details