Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 154245921-154246194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT13
Entrez Gene ID
114805
Gene Name
polypeptide N-acetylgalactosaminyltransferase 13
Gene Aliases
GalNAc-T13
UniGene
Hs.470277
Gene Chromosome Location
Chr.2: 153871898-154454177 on Build GRCh38
Transcripts
NM_001301627.1 NM_052917.3 XM_011510537.2 XM_011510538.2 XM_017003258.1 XM_017003259.1 XM_017003260.1 XM_017003261.1 XM_017003262.1 XM_017003263.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4582720 c.846A>G p.T282T Chr.2: 154245971-154245971 on Build GRCh38
3299319 c.823G>T p.D275Y Chr.2: 154245948-154245948 on Build GRCh38
4624438 c.856A>G p.R286G Chr.2: 154245981-154245981 on Build GRCh38
4893854 c.802C>T p.P268S Chr.2: 154245927-154245927 on Build GRCh38
5017774 c.808delC p.Q271fs*35 Chr.2: 154245933-154245933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11689477 Chr.2: 154245997-154245997 on Build GRCh38
rs757620709 Chr.2: 154246024-154246024 on Build GRCh38
rs773244079 Chr.2: 154245994-154245994 on Build GRCh38
rs565300387 Chr.2: 154246099-154246099 on Build GRCh38
rs542611141 Chr.2: 154246015-154246015 on Build GRCh38
rs765655215 Chr.2: 154246025-154246025 on Build GRCh38
rs752189198 Chr.2: 154246012-154246012 on Build GRCh38
rs747085291 Chr.2: 154245991-154245991 on Build GRCh38
rs754288330 Chr.2: 154246020-154246020 on Build GRCh38
rs1023184781 Chr.2: 154245989-154245989 on Build GRCh38
rs918805619 Chr.2: 154246135-154246135 on Build GRCh38
rs770790426 Chr.2: 154245998-154245998 on Build GRCh38
rs370915252 Chr.2: 154246030-154246030 on Build GRCh38
rs973329344 Chr.2: 154246106-154246106 on Build GRCh38
rs759170858 Chr.2: 154246000-154246000 on Build GRCh38
rs746008260 Chr.2: 154245978-154245978 on Build GRCh38
rs757571713 Chr.2: 154245970-154245970 on Build GRCh38
rs533695762 Chr.2: 154245985-154245985 on Build GRCh38
rs573445109 Chr.2: 154246060-154246060 on Build GRCh38
rs962821394 Chr.2: 154246075-154246075 on Build GRCh38
rs556946096 Chr.2: 154245986-154245986 on Build GRCh38
rs116692653 Chr.2: 154246027-154246027 on Build GRCh38
rs769891991 Chr.2: 154245992-154245992 on Build GRCh38
rs774285023 Chr.2: 154245999-154245999 on Build GRCh38
rs767176614 Chr.2: 154246010-154246010 on Build GRCh38
rs545613854 Chr.2: 154246097-154246097 on Build GRCh38
rs779124626 Chr.2: 154245971-154245971 on Build GRCh38
rs764609565 Chr.2: 154246019-154246019 on Build GRCh38
rs754236942 Chr.2: 154245967-154245967 on Build GRCh38

View Full Product Details