Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53650698-53650971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1D
Entrez Gene ID
776
Gene Name
calcium voltage-gated channel subunit alpha1 D
Gene Aliases
CACH3, CACN4, CACNL1A2, CCHL1A2, Cav1.3, PASNA, SANDD
UniGene
Hs.476358
Gene Chromosome Location
Chr.3: 53495049-53813151 on Build GRCh38
Transcripts
NM_000720.3 NM_001128839.2 NM_001128840.2 XM_005265448.3 XM_011534094.2 XM_011534096.2 XM_011534097.2 XM_011534100.2 XM_017007137.1 XM_017007138.1 XM_017007139.1 XM_017007140.1 XM_017007141.1 XM_017007142.1 XM_017007143.1 XM_017007144.1 XM_017007145.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3916432 c.592A>G p.N198D Chr.3: 53650887-53650887 on Build GRCh38
5693679 c.548A>T p.Y183F Chr.3: 53650843-53650843 on Build GRCh38
2780715 c.522C>T p.V174V Chr.3: 53650817-53650817 on Build GRCh38
5816561 c.622G>A p.G208R Chr.3: 53650917-53650917 on Build GRCh38
1424643 c.577G>C p.V193L Chr.3: 53650872-53650872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768486970 Chr.3: 53650741-53650741 on Build GRCh38
rs963582380 Chr.3: 53650826-53650826 on Build GRCh38
rs765547601 Chr.3: 53650871-53650871 on Build GRCh38
rs752051418 Chr.3: 53650935-53650935 on Build GRCh38
rs758468985 Chr.3: 53650928-53650928 on Build GRCh38
rs370962197 Chr.3: 53650752-53650752 on Build GRCh38
rs201812661 Chr.3: 53650905-53650905 on Build GRCh38
rs577470690 Chr.3: 53650884-53650884 on Build GRCh38
rs927953322 Chr.3: 53650811-53650811 on Build GRCh38
rs1015540260 Chr.3: 53650756-53650756 on Build GRCh38
rs989787642 Chr.3: 53650760-53650760 on Build GRCh38
rs376541461 Chr.3: 53650740-53650740 on Build GRCh38
rs1042358908 Chr.3: 53650802-53650802 on Build GRCh38
rs781685289 Chr.3: 53650948-53650948 on Build GRCh38
rs200059470 Chr.3: 53650823-53650823 on Build GRCh38
rs773262793 Chr.3: 53650930-53650930 on Build GRCh38
rs762353490 Chr.3: 53650932-53650932 on Build GRCh38
rs761433035 Chr.3: 53650762-53650762 on Build GRCh38
rs202147000 Chr.3: 53650936-53650936 on Build GRCh38
rs111366170 Chr.3: 53650841-53650841 on Build GRCh38
rs760620751 Chr.3: 53650840-53650840 on Build GRCh38
rs771872770 Chr.3: 53650813-53650813 on Build GRCh38
rs994849125 Chr.3: 53650842-53650842 on Build GRCh38
rs547551208 Chr.3: 53650865-53650865 on Build GRCh38
rs752689987 Chr.3: 53650889-53650889 on Build GRCh38
rs759742147 Chr.3: 53650853-53650853 on Build GRCh38
rs915202843 Chr.3: 53650761-53650761 on Build GRCh38
rs773161211 Chr.3: 53650817-53650817 on Build GRCh38

View Full Product Details