Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20032255-20032520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR139
Entrez Gene ID
124274
Gene Name
G protein-coupled receptor 139
Gene Aliases
GPRg1, PGR3
UniGene
Hs.446875
Gene Chromosome Location
Chr.16: 20031485-20073778 on Build GRCh38
Transcripts
NM_001002911.3 NM_001318483.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6673743 c.385G>A p.A129T Chr.16: 20032412-20032412 on Build GRCh38
3506842 c.519C>A p.D173E Chr.16: 20032278-20032278 on Build GRCh38
6838990 c.377G>C p.R126T Chr.16: 20032420-20032420 on Build GRCh38
3506843 c.514G>A p.E172K Chr.16: 20032283-20032283 on Build GRCh38
3888048 c.377G>A p.R126K Chr.16: 20032420-20032420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764757888 Chr.16: 20032437-20032437 on Build GRCh38
rs775925795 Chr.16: 20032413-20032413 on Build GRCh38
rs1039722278 Chr.16: 20032311-20032311 on Build GRCh38
rs761008511 Chr.16: 20032416-20032416 on Build GRCh38
rs778511361 Chr.16: 20032366-20032366 on Build GRCh38
rs537730436 Chr.16: 20032375-20032375 on Build GRCh38
rs779648316 Chr.16: 20032479-20032479 on Build GRCh38
rs141137866 Chr.16: 20032284-20032284 on Build GRCh38
rs758080532 Chr.16: 20032449-20032449 on Build GRCh38
rs549998420 Chr.16: 20032383-20032383 on Build GRCh38
rs571459092 Chr.16: 20032392-20032392 on Build GRCh38
rs779166553 Chr.16: 20032350-20032350 on Build GRCh38
rs753339106 Chr.16: 20032353-20032353 on Build GRCh38
rs756813774 Chr.16: 20032360-20032360 on Build GRCh38
rs770710839 Chr.16: 20032451-20032451 on Build GRCh38
rs868806533 Chr.16: 20032288-20032288 on Build GRCh38
rs117796776 Chr.16: 20032364-20032364 on Build GRCh38
rs371060930 Chr.16: 20032351-20032351 on Build GRCh38
rs751365983 Chr.16: 20032481-20032481 on Build GRCh38
rs777131487 Chr.16: 20032380-20032380 on Build GRCh38
rs890668532 Chr.16: 20032275-20032276 on Build GRCh38
rs749993997 Chr.16: 20032439-20032439 on Build GRCh38
rs1013940734 Chr.16: 20032330-20032330 on Build GRCh38
rs767966063 Chr.16: 20032316-20032316 on Build GRCh38
rs975483612 Chr.16: 20032425-20032425 on Build GRCh38
rs142100378 Chr.16: 20032426-20032426 on Build GRCh38
rs750115024 Chr.16: 20032367-20032367 on Build GRCh38
rs1015377222 Chr.16: 20032456-20032456 on Build GRCh38
rs759836680 Chr.16: 20032399-20032399 on Build GRCh38
rs146922265 Chr.16: 20032384-20032384 on Build GRCh38
rs748486913 Chr.16: 20032434-20032434 on Build GRCh38
rs758087867 Chr.16: 20032370-20032370 on Build GRCh38
rs754787961 Chr.16: 20032482-20032482 on Build GRCh38
rs895468316 Chr.16: 20032320-20032320 on Build GRCh38
rs753106637 Chr.16: 20032345-20032345 on Build GRCh38
rs756668514 Chr.16: 20032346-20032346 on Build GRCh38
rs761281280 Chr.16: 20032435-20032435 on Build GRCh38
rs137883582 Chr.16: 20032386-20032386 on Build GRCh38
rs1009146155 Chr.16: 20032301-20032301 on Build GRCh38
rs1003961744 Chr.16: 20032446-20032446 on Build GRCh38
rs538799671 Chr.16: 20032429-20032429 on Build GRCh38
rs1032845174 Chr.16: 20032412-20032412 on Build GRCh38
rs371518776 Chr.16: 20032398-20032398 on Build GRCh38
rs748140778 Chr.16: 20032496-20032496 on Build GRCh38
rs1000398834 Chr.16: 20032361-20032361 on Build GRCh38
rs982809076 Chr.16: 20032497-20032497 on Build GRCh38
rs759798192 Chr.16: 20032305-20032305 on Build GRCh38
rs1001230865 Chr.16: 20032276-20032276 on Build GRCh38
rs770936651 Chr.16: 20032400-20032400 on Build GRCh38
rs749366228 Chr.16: 20032385-20032385 on Build GRCh38
rs781252777 Chr.16: 20032486-20032486 on Build GRCh38

View Full Product Details