Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 68526146-68526373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APLF
Entrez Gene ID
200558
Gene Name
aprataxin and PNKP like factor
Gene Aliases
APFL, C2orf13, PALF, Xip1, ZCCHH1
UniGene
Hs.720369
Gene Chromosome Location
Chr.2: 68467559-68580162 on Build GRCh38
Transcripts
NM_173545.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4598649 c.716C>T p.S239L Chr.2: 68526154-68526154 on Build GRCh38
5717022 c.797C>T p.S266L Chr.2: 68526235-68526235 on Build GRCh38
3055677 c.755G>T p.G252V Chr.2: 68526193-68526193 on Build GRCh38
5758549 c.753A>G p.T251T Chr.2: 68526191-68526191 on Build GRCh38
4831411 c.727G>A p.E243K Chr.2: 68526165-68526165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777116731 Chr.2: 68526201-68526201 on Build GRCh38
rs952803388 Chr.2: 68526236-68526236 on Build GRCh38
rs926032857 Chr.2: 68526308-68526308 on Build GRCh38
rs149184095 Chr.2: 68526228-68526228 on Build GRCh38
rs774190469 Chr.2: 68526271-68526272 on Build GRCh38
rs765930328 Chr.2: 68526198-68526198 on Build GRCh38
rs753165035 Chr.2: 68526224-68526224 on Build GRCh38
rs547819103 Chr.2: 68526344-68526344 on Build GRCh38
rs749158306 Chr.2: 68526281-68526281 on Build GRCh38
rs759081097 Chr.2: 68526284-68526285 on Build GRCh38
rs781240968 Chr.2: 68526257-68526257 on Build GRCh38
rs1033313323 Chr.2: 68526250-68526250 on Build GRCh38
rs751629538 Chr.2: 68526247-68526247 on Build GRCh38
rs779023996 Chr.2: 68526292-68526292 on Build GRCh38
rs1025010079 Chr.2: 68526261-68526261 on Build GRCh38
rs957451770 Chr.2: 68526332-68526332 on Build GRCh38
rs898257933 Chr.2: 68526197-68526197 on Build GRCh38
rs754681879 Chr.2: 68526284-68526284 on Build GRCh38
rs555965632 Chr.2: 68526289-68526290 on Build GRCh38
rs766105884 Chr.2: 68526271-68526271 on Build GRCh38
rs200252250 Chr.2: 68526297-68526298 on Build GRCh38
rs527990506 Chr.2: 68526203-68526203 on Build GRCh38
rs758480117 Chr.2: 68526225-68526225 on Build GRCh38
rs762551893 Chr.2: 68526191-68526191 on Build GRCh38
rs765819873 Chr.2: 68526204-68526204 on Build GRCh38
rs548669995 Chr.2: 68526251-68526251 on Build GRCh38

View Full Product Details