Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 34524592-34524857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EAPP
Entrez Gene ID
55837
Gene Name
E2F associated phosphoprotein
Gene Aliases
BM036, C14orf11
UniGene
Hs.433269
Gene Chromosome Location
Chr.14: 34515929-34539737 on Build GRCh38
Transcripts
NM_001318916.1 NM_018453.3 XM_011536958.1 XM_017021460.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6651109 c.518C>T p.P173L Chr.14: 34524760-34524760 on Build GRCh38
2195817 c.500G>A p.R167H Chr.14: 34524778-34524778 on Build GRCh38
1493282 c.471-2A>T p.? Chr.14: 34524809-34524809 on Build GRCh38
6651110 c.545C>T p.P182L Chr.14: 34524733-34524733 on Build GRCh38
1607637 c.549C>A p.A183A Chr.14: 34524729-34524729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745982195 Chr.14: 34524675-34524676 on Build GRCh38
rs773515055 Chr.14: 34524693-34524693 on Build GRCh38
rs752378509 Chr.14: 34524734-34524734 on Build GRCh38
rs371845934 Chr.14: 34524691-34524691 on Build GRCh38
rs772618044 Chr.14: 34524616-34524617 on Build GRCh38
rs757536810 Chr.14: 34524673-34524674 on Build GRCh38
rs755893563 Chr.14: 34524757-34524757 on Build GRCh38
rs367795110 Chr.14: 34524657-34524658 on Build GRCh38
rs768420208 Chr.14: 34524682-34524685 on Build GRCh38
rs373317079 Chr.14: 34524711-34524711 on Build GRCh38
rs778166030 Chr.14: 34524659-34524660 on Build GRCh38
rs111420332 Chr.14: 34524653-34524654 on Build GRCh38
rs61981524 Chr.14: 34524657-34524657 on Build GRCh38
rs769159699 Chr.14: 34524651-34524652 on Build GRCh38
rs778915112 Chr.14: 34524683-34524683 on Build GRCh38
rs767180036 Chr.14: 34524721-34524721 on Build GRCh38
rs71121205 Chr.14: 34524682-34524683 on Build GRCh38
rs753511351 Chr.14: 34524761-34524761 on Build GRCh38
rs748364842 Chr.14: 34524791-34524791 on Build GRCh38
rs897964084 Chr.14: 34524639-34524639 on Build GRCh38
rs747507808 Chr.14: 34524770-34524770 on Build GRCh38
rs1038376335 Chr.14: 34524630-34524630 on Build GRCh38
rs78060227 Chr.14: 34524617-34524617 on Build GRCh38
rs747005660 Chr.14: 34524684-34524684 on Build GRCh38
rs374847059 Chr.14: 34524692-34524692 on Build GRCh38
rs780415266 Chr.14: 34524755-34524755 on Build GRCh38
rs910197339 Chr.14: 34524817-34524817 on Build GRCh38
rs370105965 Chr.14: 34524710-34524710 on Build GRCh38
rs763376031 Chr.14: 34524701-34524701 on Build GRCh38
rs17352411 Chr.14: 34524776-34524776 on Build GRCh38
rs781306099 Chr.14: 34524779-34524779 on Build GRCh38
rs776801749 Chr.14: 34524685-34524685 on Build GRCh38
rs979389885 Chr.14: 34524659-34524659 on Build GRCh38
rs201898193 Chr.14: 34524656-34524657 on Build GRCh38
rs199911091 Chr.14: 34524673-34524673 on Build GRCh38
rs749487302 Chr.14: 34524671-34524672 on Build GRCh38
rs751114532 Chr.14: 34524652-34524655 on Build GRCh38
rs751450866 Chr.14: 34524653-34524653 on Build GRCh38
rs527718120 Chr.14: 34524827-34524827 on Build GRCh38
rs763980359 Chr.14: 34524760-34524760 on Build GRCh38
rs769690837 Chr.14: 34524713-34524714 on Build GRCh38
rs1011464565 Chr.14: 34524702-34524702 on Build GRCh38
rs199731169 Chr.14: 34524778-34524778 on Build GRCh38
rs757088607 Chr.14: 34524767-34524767 on Build GRCh38

View Full Product Details