Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157124969-157125234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV3
Entrez Gene ID
2117
Gene Name
ETS variant 3
Gene Aliases
METS, PE-1, PE1, bA110J1.4
UniGene
Hs.105636
Gene Chromosome Location
Chr.1: 157121191-157138591 on Build GRCh38
Transcripts
NM_001145312.1 XM_006711210.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4436126 c.1195G>T p.E399* Chr.1: 157125185-157125185 on Build GRCh38
5420770 c.1239_1240delGA p.R413fs*3 Chr.1: 157125140-157125141 on Build GRCh38
7314589 c.1377C>A p.P459P Chr.1: 157125003-157125003 on Build GRCh38
6507911 c.1345G>T p.E449* Chr.1: 157125035-157125035 on Build GRCh38
5518207 c.1198_1200delAAG p.K400delK Chr.1: 157125180-157125182 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371357999 Chr.1: 157125032-157125032 on Build GRCh38
rs372706068 Chr.1: 157125211-157125211 on Build GRCh38
rs1034429249 Chr.1: 157125187-157125187 on Build GRCh38
rs753974294 Chr.1: 157125144-157125144 on Build GRCh38
rs751272378 Chr.1: 157125076-157125076 on Build GRCh38
rs750434785 Chr.1: 157125006-157125006 on Build GRCh38
rs907823852 Chr.1: 157125072-157125072 on Build GRCh38
rs948139048 Chr.1: 157125129-157125129 on Build GRCh38
rs200902536 Chr.1: 157125010-157125010 on Build GRCh38
rs907180052 Chr.1: 157125153-157125153 on Build GRCh38
rs1025456826 Chr.1: 157125204-157125204 on Build GRCh38
rs757475929 Chr.1: 157125148-157125148 on Build GRCh38
rs778086102 Chr.1: 157125139-157125139 on Build GRCh38
rs892030089 Chr.1: 157125026-157125026 on Build GRCh38
rs1002029287 Chr.1: 157125174-157125174 on Build GRCh38
rs903862826 Chr.1: 157125160-157125160 on Build GRCh38
rs1056341464 Chr.1: 157125108-157125108 on Build GRCh38
rs983381126 Chr.1: 157125049-157125049 on Build GRCh38
rs115176550 Chr.1: 157125123-157125123 on Build GRCh38
rs867960541 Chr.1: 157125120-157125120 on Build GRCh38
rs1055144689 Chr.1: 157125019-157125019 on Build GRCh38
rs766238220 Chr.1: 157125038-157125038 on Build GRCh38
rs898349818 Chr.1: 157125202-157125202 on Build GRCh38
rs778841429 Chr.1: 157125170-157125170 on Build GRCh38
rs746432551 Chr.1: 157125192-157125192 on Build GRCh38
rs540027756 Chr.1: 157125106-157125106 on Build GRCh38
rs545598508 Chr.1: 157125203-157125203 on Build GRCh38
rs939180582 Chr.1: 157125077-157125077 on Build GRCh38

View Full Product Details