Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44213687-44213932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMAP1
Entrez Gene ID
55929
Gene Name
DNA methyltransferase 1 associated protein 1
Gene Aliases
DNMAP1, DNMTAP1, EAF2, MEAF2, SWC4
UniGene
Hs.8008
Gene Chromosome Location
Chr.1: 44213428-44220679 on Build GRCh38
Transcripts
NM_001034023.1 NM_001034024.1 NM_019100.4 XM_005271037.4 XM_005271039.1 XM_005271040.1 XM_011541784.2 XM_011541785.2 XM_017001807.1 XM_017001808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7195906 c.57T>C p.D19D Chr.1: 44213810-44213810 on Build GRCh38
6212583 c.96delC p.P33fs*13 Chr.1: 44213849-44213849 on Build GRCh38
6212584 c.96delC p.P33fs*13 Chr.1: 44213849-44213849 on Build GRCh38
6560898 c.55G>A p.D19N Chr.1: 44213808-44213808 on Build GRCh38
7195907 c.57T>C p.D19D Chr.1: 44213810-44213810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532299749 Chr.1: 44213909-44213909 on Build GRCh38
rs762185364 Chr.1: 44213762-44213762 on Build GRCh38
rs148699903 Chr.1: 44213894-44213894 on Build GRCh38
rs189226255 Chr.1: 44213857-44213857 on Build GRCh38
rs949855912 Chr.1: 44213900-44213904 on Build GRCh38
rs921702625 Chr.1: 44213740-44213740 on Build GRCh38
rs777534450 Chr.1: 44213722-44213722 on Build GRCh38
rs762931520 Chr.1: 44213833-44213833 on Build GRCh38
rs777283361 Chr.1: 44213891-44213891 on Build GRCh38
rs563212663 Chr.1: 44213908-44213908 on Build GRCh38
rs761109455 Chr.1: 44213745-44213745 on Build GRCh38
rs868865806 Chr.1: 44213851-44213851 on Build GRCh38
rs750725590 Chr.1: 44213825-44213825 on Build GRCh38
rs776941173 Chr.1: 44213752-44213752 on Build GRCh38
rs866800708 Chr.1: 44213805-44213805 on Build GRCh38
rs775419996 Chr.1: 44213898-44213898 on Build GRCh38
rs769155234 Chr.1: 44213750-44213750 on Build GRCh38
rs917178059 Chr.1: 44213899-44213899 on Build GRCh38
rs867078728 Chr.1: 44213794-44213794 on Build GRCh38
rs754930307 Chr.1: 44213871-44213871 on Build GRCh38
rs552131585 Chr.1: 44213914-44213914 on Build GRCh38
rs1028034898 Chr.1: 44213793-44213793 on Build GRCh38
rs770538505 Chr.1: 44213725-44213725 on Build GRCh38
rs756016420 Chr.1: 44213887-44213887 on Build GRCh38
rs765353094 Chr.1: 44213812-44213812 on Build GRCh38
rs747991748 Chr.1: 44213721-44213721 on Build GRCh38
rs748712689 Chr.1: 44213724-44213724 on Build GRCh38
rs751694516 Chr.1: 44213867-44213867 on Build GRCh38
rs374557407 Chr.1: 44213757-44213757 on Build GRCh38
rs766424456 Chr.1: 44213838-44213838 on Build GRCh38
rs371549437 Chr.1: 44213886-44213886 on Build GRCh38
rs803369 Chr.1: 44213884-44213884 on Build GRCh38
rs775842817 Chr.1: 44213737-44213737 on Build GRCh38

View Full Product Details