Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110627376-110627611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAXD
Entrez Gene ID
55739
Gene Name
NAD(P)HX dehydratase
Gene Aliases
CARKD, LP3298
UniGene
Hs.408324
Gene Chromosome Location
Chr.13: 110615460-110639996 on Build GRCh38
Transcripts
NM_001242881.1 NM_001242882.1 NM_001242883.1 NM_018210.3 NR_040103.1 NR_040104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
344238 c.414G>A p.V138V Chr.13: 110627466-110627466 on Build GRCh38
4611505 c.445_447delGTC p.V150delV Chr.13: 110627497-110627499 on Build GRCh38
945385 c.404T>C p.V135A Chr.13: 110627456-110627456 on Build GRCh38
6402970 c.449T>C p.V150A Chr.13: 110627501-110627501 on Build GRCh38
7271627 c.417G>A p.E139E Chr.13: 110627469-110627469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766274714 Chr.13: 110627467-110627467 on Build GRCh38
rs1044112 Chr.13: 110627506-110627506 on Build GRCh38
rs749047531 Chr.13: 110627589-110627589 on Build GRCh38
rs750729449 Chr.13: 110627502-110627502 on Build GRCh38
rs757598469 Chr.13: 110627426-110627426 on Build GRCh38
rs767422095 Chr.13: 110627479-110627479 on Build GRCh38
rs779315237 Chr.13: 110627432-110627432 on Build GRCh38
rs543549091 Chr.13: 110627581-110627581 on Build GRCh38
rs3742192 Chr.13: 110627497-110627497 on Build GRCh38
rs866367234 Chr.13: 110627462-110627462 on Build GRCh38
rs1044130080 Chr.13: 110627555-110627555 on Build GRCh38
rs41306678 Chr.13: 110627500-110627500 on Build GRCh38
rs553217045 Chr.13: 110627469-110627469 on Build GRCh38
rs749538253 Chr.13: 110627425-110627425 on Build GRCh38
rs759336239 Chr.13: 110627562-110627562 on Build GRCh38
rs964362259 Chr.13: 110627488-110627488 on Build GRCh38
rs377595355 Chr.13: 110627454-110627454 on Build GRCh38
rs764210701 Chr.13: 110627490-110627490 on Build GRCh38
rs371103258 Chr.13: 110627552-110627552 on Build GRCh38
rs764143271 Chr.13: 110627579-110627579 on Build GRCh38
rs3742191 Chr.13: 110627470-110627470 on Build GRCh38
rs373944589 Chr.13: 110627580-110627580 on Build GRCh38
rs976332651 Chr.13: 110627478-110627478 on Build GRCh38
rs758717285 Chr.13: 110627516-110627516 on Build GRCh38
rs756157326 Chr.13: 110627413-110627413 on Build GRCh38
rs771788978 Chr.13: 110627566-110627566 on Build GRCh38
rs746345462 Chr.13: 110627444-110627444 on Build GRCh38
rs377430322 Chr.13: 110627521-110627521 on Build GRCh38
rs757530909 Chr.13: 110627499-110627499 on Build GRCh38
rs55839400 Chr.13: 110627450-110627450 on Build GRCh38
rs373714648 Chr.13: 110627451-110627451 on Build GRCh38
rs777934932 Chr.13: 110627415-110627415 on Build GRCh38
rs1056811855 Chr.13: 110627468-110627468 on Build GRCh38
rs534898507 Chr.13: 110627459-110627459 on Build GRCh38
rs576045093 Chr.13: 110627528-110627528 on Build GRCh38
rs775478477 Chr.13: 110627568-110627568 on Build GRCh38
rs201831524 Chr.13: 110627483-110627483 on Build GRCh38
rs1048872804 Chr.13: 110627472-110627472 on Build GRCh38
rs773880857 Chr.13: 110627557-110627557 on Build GRCh38
rs928388068 Chr.13: 110627571-110627571 on Build GRCh38
rs762733351 Chr.13: 110627460-110627460 on Build GRCh38
rs777490219 Chr.13: 110627529-110627529 on Build GRCh38
rs146504568 Chr.13: 110627482-110627482 on Build GRCh38
rs888537853 Chr.13: 110627476-110627476 on Build GRCh38
rs760647801 Chr.13: 110627575-110627575 on Build GRCh38

View Full Product Details