Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66858335-66858584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRFN4
Entrez Gene ID
78999
Gene Name
leucine rich repeat and fibronectin type III domain containing 4
Gene Aliases
FIGLER6, SALM3, SALM3.
UniGene
Hs.209979
Gene Chromosome Location
Chr.11: 66857087-66860475 on Build GRCh38
Transcripts
NM_024036.4 XM_005274239.3 NM_000920.3 NM_001040716.1 NM_022172.2 XM_005274031.4 XM_005274032.4 XM_006718578.3 XM_011545086.2 XM_017017868.1 XM_017017869.1 XM_017017870.1 XM_017017871.1 XM_017017872.1

Gene Symbol
PC
Entrez Gene ID
5091
Gene Name
pyruvate carboxylase
Gene Aliases
PCB
UniGene
Hs.89890
Gene Chromosome Location
Chr.11: 66848522-66958418 on Build GRCh38
Transcripts
NM_024036.4 XM_005274239.3 NM_000920.3 NM_001040716.1 NM_022172.2 XM_005274031.4 XM_005274032.4 XM_006718578.3 XM_011545086.2 XM_017017868.1 XM_017017869.1 XM_017017870.1 XM_017017871.1 XM_017017872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4649477 c.816C>T p.P272P Chr.11: 66858560-66858560 on Build GRCh38
4638845 c.809C>T p.A270V Chr.11: 66858553-66858553 on Build GRCh38
4698711 c.616G>A p.A206T Chr.11: 66858360-66858360 on Build GRCh38
5037048 c.689G>A p.S230N Chr.11: 66858433-66858433 on Build GRCh38
6501439 c.661G>C p.E221Q Chr.11: 66858405-66858405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs908758854 Chr.11: 66858554-66858554 on Build GRCh38
rs552808171 Chr.11: 66858498-66858498 on Build GRCh38
rs754636227 Chr.11: 66858526-66858526 on Build GRCh38
rs746326458 Chr.11: 66858423-66858423 on Build GRCh38
rs377099719 Chr.11: 66858359-66858359 on Build GRCh38
rs763082444 Chr.11: 66858496-66858496 on Build GRCh38
rs747655680 Chr.11: 66858529-66858529 on Build GRCh38
rs35165280 Chr.11: 66858440-66858441 on Build GRCh38
rs770206065 Chr.11: 66858560-66858560 on Build GRCh38
rs750352828 Chr.11: 66858503-66858503 on Build GRCh38
rs762107008 Chr.11: 66858512-66858512 on Build GRCh38
rs748957599 Chr.11: 66858416-66858416 on Build GRCh38
rs893634067 Chr.11: 66858388-66858388 on Build GRCh38
rs983603393 Chr.11: 66858405-66858405 on Build GRCh38
rs781174132 Chr.11: 66858386-66858386 on Build GRCh38
rs754095341 Chr.11: 66858365-66858365 on Build GRCh38
rs906130722 Chr.11: 66858524-66858524 on Build GRCh38
rs184634237 Chr.11: 66858536-66858536 on Build GRCh38
rs759967981 Chr.11: 66858546-66858546 on Build GRCh38
rs539362583 Chr.11: 66858491-66858491 on Build GRCh38
rs904516692 Chr.11: 66858470-66858470 on Build GRCh38
rs138224455 Chr.11: 66858541-66858541 on Build GRCh38
rs771591285 Chr.11: 66858533-66858533 on Build GRCh38
rs755206334 Chr.11: 66858368-66858368 on Build GRCh38
rs755906892 Chr.11: 66858396-66858396 on Build GRCh38
rs948508886 Chr.11: 66858519-66858519 on Build GRCh38
rs752758622 Chr.11: 66858379-66858379 on Build GRCh38
rs867603133 Chr.11: 66858441-66858441 on Build GRCh38
rs755956386 Chr.11: 66858506-66858506 on Build GRCh38
rs1010816428 Chr.11: 66858417-66858417 on Build GRCh38
rs767393572 Chr.11: 66858497-66858497 on Build GRCh38
rs761099728 Chr.11: 66858355-66858355 on Build GRCh38
rs775748145 Chr.11: 66858564-66858564 on Build GRCh38
rs771448599 Chr.11: 66858481-66858481 on Build GRCh38
rs757156530 Chr.11: 66858380-66858380 on Build GRCh38
rs773740511 Chr.11: 66858435-66858435 on Build GRCh38
rs35367010 Chr.11: 66858446-66858447 on Build GRCh38
rs946577439 Chr.11: 66858429-66858429 on Build GRCh38
rs779730278 Chr.11: 66858401-66858401 on Build GRCh38
rs200279686 Chr.11: 66858515-66858515 on Build GRCh38
rs760998247 Chr.11: 66858461-66858461 on Build GRCh38
rs370566594 Chr.11: 66858422-66858422 on Build GRCh38
rs367553596 Chr.11: 66858421-66858421 on Build GRCh38
rs762991561 Chr.11: 66858566-66858566 on Build GRCh38
rs866286266 Chr.11: 66858467-66858467 on Build GRCh38
rs777097562 Chr.11: 66858489-66858489 on Build GRCh38
rs537084771 Chr.11: 66858373-66858373 on Build GRCh38
rs558719118 Chr.11: 66858410-66858410 on Build GRCh38
rs369728361 Chr.11: 66858402-66858402 on Build GRCh38
rs550930114 Chr.11: 66858395-66858395 on Build GRCh38
rs765499702 Chr.11: 66858490-66858490 on Build GRCh38
rs535268151 Chr.11: 66858528-66858528 on Build GRCh38
rs961419932 Chr.11: 66858563-66858563 on Build GRCh38
rs983248854 Chr.11: 66858557-66858557 on Build GRCh38
rs866591797 Chr.11: 66858468-66858468 on Build GRCh38
rs921863705 Chr.11: 66858428-66858428 on Build GRCh38
rs771499528 Chr.11: 66858540-66858540 on Build GRCh38

View Full Product Details