Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 27780797-27781048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL42
Entrez Gene ID
57542
Gene Name
kelch like family member 42
Gene Aliases
Ctb9, KLHDC5
UniGene
Hs.505104
Gene Chromosome Location
Chr.12: 27779957-27803040 on Build GRCh38
Transcripts
NM_020782.1 XM_017019698.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6309663 c.559G>T p.E187* Chr.12: 27780889-27780889 on Build GRCh38
7181251 c.654C>T p.S218S Chr.12: 27780984-27780984 on Build GRCh38
6696839 c.505C>A p.L169M Chr.12: 27780835-27780835 on Build GRCh38
7174136 c.643G>A p.E215K Chr.12: 27780973-27780973 on Build GRCh38
4641684 c.485G>A p.C162Y Chr.12: 27780815-27780815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763249165 Chr.12: 27780992-27780992 on Build GRCh38
rs571539832 Chr.12: 27780903-27780903 on Build GRCh38
rs750629733 Chr.12: 27780865-27780865 on Build GRCh38
rs984170691 Chr.12: 27780976-27780976 on Build GRCh38
rs202013500 Chr.12: 27780818-27780818 on Build GRCh38
rs143561055 Chr.12: 27780822-27780822 on Build GRCh38
rs764503221 Chr.12: 27780997-27780997 on Build GRCh38
rs1057131661 Chr.12: 27781023-27781023 on Build GRCh38
rs983596535 Chr.12: 27780889-27780889 on Build GRCh38
rs757348968 Chr.12: 27780854-27780854 on Build GRCh38
rs891730898 Chr.12: 27781024-27781024 on Build GRCh38
rs774837373 Chr.12: 27781013-27781013 on Build GRCh38
rs776041154 Chr.12: 27780939-27780939 on Build GRCh38
rs139400429 Chr.12: 27780918-27780918 on Build GRCh38
rs551232636 Chr.12: 27780937-27780937 on Build GRCh38
rs777794545 Chr.12: 27780924-27780924 on Build GRCh38
rs762267669 Chr.12: 27781015-27781015 on Build GRCh38
rs146766343 Chr.12: 27780902-27780902 on Build GRCh38
rs1018542224 Chr.12: 27780843-27780843 on Build GRCh38
rs746975250 Chr.12: 27780925-27780925 on Build GRCh38
rs868098269 Chr.12: 27780896-27780896 on Build GRCh38
rs747911594 Chr.12: 27780908-27780908 on Build GRCh38
rs965994775 Chr.12: 27780862-27780862 on Build GRCh38
rs764521544 Chr.12: 27780848-27780848 on Build GRCh38
rs769277069 Chr.12: 27780963-27780963 on Build GRCh38
rs146472087 Chr.12: 27780990-27780990 on Build GRCh38
rs756341324 Chr.12: 27780871-27780871 on Build GRCh38
rs929533960 Chr.12: 27780878-27780878 on Build GRCh38
rs745508584 Chr.12: 27780948-27780948 on Build GRCh38
rs750421038 Chr.12: 27781029-27781029 on Build GRCh38
rs916527623 Chr.12: 27781028-27781028 on Build GRCh38
rs763342736 Chr.12: 27780836-27780836 on Build GRCh38
rs767613843 Chr.12: 27781020-27781020 on Build GRCh38
rs767561636 Chr.12: 27780863-27780863 on Build GRCh38
rs937353248 Chr.12: 27781009-27781009 on Build GRCh38
rs991883912 Chr.12: 27781011-27781011 on Build GRCh38

View Full Product Details