Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54252295-54252552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LILRB5
Entrez Gene ID
10990
Gene Name
leukocyte immunoglobulin like receptor B5
Gene Aliases
CD85C, LIR-8, LIR8
UniGene
Hs.655338
Gene Chromosome Location
Chr.19: 54250405-54257308 on Build GRCh38
Transcripts
NM_001081442.2 NM_001081443.2 NM_001304457.1 NM_006840.4 XM_011526359.2 XM_011526360.2 XM_011526361.2 XM_011526362.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4132483 c.1500G>A p.A500A Chr.19: 54252521-54252521 on Build GRCh38
5028623 c.1473C>G p.A491A Chr.19: 54252548-54252548 on Build GRCh38
4418691 c.1573+8A>G p.? Chr.19: 54252358-54252358 on Build GRCh38
5056920 c.1523G>T p.G508V Chr.19: 54252498-54252498 on Build GRCh38
170869 c.1484G>A p.R495H Chr.19: 54252537-54252537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762519162 Chr.19: 54252522-54252522 on Build GRCh38
rs768252674 Chr.19: 54252437-54252437 on Build GRCh38
rs3859536 Chr.19: 54252324-54252324 on Build GRCh38
rs781013496 Chr.19: 54252454-54252454 on Build GRCh38
rs1043921231 Chr.19: 54252525-54252525 on Build GRCh38
rs558957309 Chr.19: 54252411-54252411 on Build GRCh38
rs764856929 Chr.19: 54252519-54252519 on Build GRCh38
rs200241141 Chr.19: 54252417-54252417 on Build GRCh38
rs575908444 Chr.19: 54252413-54252413 on Build GRCh38
rs769020351 Chr.19: 54252409-54252409 on Build GRCh38
rs559028253 Chr.19: 54252486-54252486 on Build GRCh38
rs745979942 Chr.19: 54252395-54252395 on Build GRCh38
rs759653236 Chr.19: 54252327-54252327 on Build GRCh38
rs764400845 Chr.19: 54252341-54252341 on Build GRCh38
rs776117393 Chr.19: 54252401-54252401 on Build GRCh38
rs916720323 Chr.19: 54252522-54252524 on Build GRCh38
rs764850667 Chr.19: 54252448-54252448 on Build GRCh38
rs34318516 Chr.19: 54252402-54252402 on Build GRCh38
rs147630506 Chr.19: 54252385-54252385 on Build GRCh38
rs745778428 Chr.19: 54252458-54252458 on Build GRCh38
rs202072198 Chr.19: 54252496-54252496 on Build GRCh38
rs750923778 Chr.19: 54252415-54252415 on Build GRCh38
rs781148904 Chr.19: 54252394-54252394 on Build GRCh38
rs774756316 Chr.19: 54252439-54252439 on Build GRCh38
rs1052261503 Chr.19: 54252489-54252489 on Build GRCh38
rs148331360 Chr.19: 54252333-54252333 on Build GRCh38
rs763884195 Chr.19: 54252526-54252526 on Build GRCh38
rs969443200 Chr.19: 54252386-54252386 on Build GRCh38
rs117421142 Chr.19: 54252383-54252383 on Build GRCh38
rs567028226 Chr.19: 54252406-54252406 on Build GRCh38
rs778483312 Chr.19: 54252365-54252365 on Build GRCh38
rs371297644 Chr.19: 54252407-54252407 on Build GRCh38
rs150565517 Chr.19: 54252379-54252379 on Build GRCh38
rs770838304 Chr.19: 54252393-54252393 on Build GRCh38
rs773627488 Chr.19: 54252506-54252506 on Build GRCh38
rs113466981 Chr.19: 54252318-54252318 on Build GRCh38
rs867811558 Chr.19: 54252516-54252516 on Build GRCh38
rs775667688 Chr.19: 54252335-54252335 on Build GRCh38
rs141451449 Chr.19: 54252350-54252350 on Build GRCh38
rs191264163 Chr.19: 54252475-54252475 on Build GRCh38
rs764766841 Chr.19: 54252418-54252418 on Build GRCh38
rs1009170253 Chr.19: 54252396-54252396 on Build GRCh38
rs140347665 Chr.19: 54252380-54252380 on Build GRCh38
rs774755963 Chr.19: 54252410-54252410 on Build GRCh38
rs757051182 Chr.19: 54252449-54252449 on Build GRCh38
rs372762503 Chr.19: 54252325-54252325 on Build GRCh38
rs415860 Chr.19: 54252439-54252439 on Build GRCh38
rs145736798 Chr.19: 54252521-54252521 on Build GRCh38
rs752063431 Chr.19: 54252349-54252349 on Build GRCh38
rs199699947 Chr.19: 54252447-54252447 on Build GRCh38
rs757033605 Chr.19: 54252532-54252532 on Build GRCh38
rs950401310 Chr.19: 54252467-54252467 on Build GRCh38
rs769766355 Chr.19: 54252461-54252461 on Build GRCh38
rs754480826 Chr.19: 54252364-54252364 on Build GRCh38
rs142288703 Chr.19: 54252505-54252505 on Build GRCh38
rs751171496 Chr.19: 54252528-54252528 on Build GRCh38
rs902481830 Chr.19: 54252370-54252370 on Build GRCh38
rs199591615 Chr.19: 54252515-54252515 on Build GRCh38
rs3843753 Chr.19: 54252358-54252358 on Build GRCh38
rs752405305 Chr.19: 54252428-54252428 on Build GRCh38
rs946990742 Chr.19: 54252524-54252524 on Build GRCh38
rs763330079 Chr.19: 54252338-54252338 on Build GRCh38
rs772437158 Chr.19: 54252502-54252502 on Build GRCh38

View Full Product Details