Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39870243-39870470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCGBP
Entrez Gene ID
8857
Gene Name
Fc fragment of IgG binding protein
Gene Aliases
FC(GAMMA)BP
UniGene
Hs.111732
Gene Chromosome Location
Chr.19: 39863323-39934626 on Build GRCh38
Transcripts

SNP Detail

SNP ID Chromosome Location
rs370106966 Chr.19: 39870398-39870398 on Build GRCh38
rs920374479 Chr.19: 39870307-39870307 on Build GRCh38
rs769584991 Chr.19: 39870317-39870317 on Build GRCh38
rs778971026 Chr.19: 39870397-39870397 on Build GRCh38
rs754164424 Chr.19: 39870339-39870339 on Build GRCh38
rs145067279 Chr.19: 39870333-39870333 on Build GRCh38
rs774301932 Chr.19: 39870266-39870266 on Build GRCh38
rs200378087 Chr.19: 39870415-39870415 on Build GRCh38
rs768745210 Chr.19: 39870404-39870404 on Build GRCh38
rs139526217 Chr.19: 39870296-39870296 on Build GRCh38
rs747851180 Chr.19: 39870305-39870305 on Build GRCh38
rs767754332 Chr.19: 39870330-39870330 on Build GRCh38
rs750884516 Chr.19: 39870427-39870427 on Build GRCh38
rs200125442 Chr.19: 39870297-39870297 on Build GRCh38
rs749023392 Chr.19: 39870383-39870383 on Build GRCh38
rs764451016 Chr.19: 39870335-39870335 on Build GRCh38
rs767840667 Chr.19: 39870271-39870271 on Build GRCh38
rs757705958 Chr.19: 39870290-39870290 on Build GRCh38
rs768835655 Chr.19: 39870420-39870421 on Build GRCh38
rs887323887 Chr.19: 39870321-39870321 on Build GRCh38
rs184463001 Chr.19: 39870334-39870334 on Build GRCh38
rs747761387 Chr.19: 39870366-39870366 on Build GRCh38
rs745822010 Chr.19: 39870401-39870401 on Build GRCh38
rs867136565 Chr.19: 39870340-39870340 on Build GRCh38
rs957673469 Chr.19: 39870369-39870369 on Build GRCh38
rs920931449 Chr.19: 39870364-39870364 on Build GRCh38
rs759774406 Chr.19: 39870269-39870269 on Build GRCh38
rs776973740 Chr.19: 39870414-39870414 on Build GRCh38
rs540284772 Chr.19: 39870402-39870402 on Build GRCh38
rs754329544 Chr.19: 39870289-39870289 on Build GRCh38
rs367934901 Chr.19: 39870373-39870373 on Build GRCh38
rs753007914 Chr.19: 39870272-39870272 on Build GRCh38
rs779557013 Chr.19: 39870293-39870293 on Build GRCh38
rs749544284 Chr.19: 39870325-39870329 on Build GRCh38
rs763485062 Chr.19: 39870448-39870448 on Build GRCh38
rs766444672 Chr.19: 39870263-39870263 on Build GRCh38
rs774405155 Chr.19: 39870325-39870325 on Build GRCh38
rs780707707 Chr.19: 39870352-39870352 on Build GRCh38
rs754466025 Chr.19: 39870351-39870351 on Build GRCh38
rs73930662 Chr.19: 39870372-39870372 on Build GRCh38
rs575240993 Chr.19: 39870320-39870320 on Build GRCh38
rs201020566 Chr.19: 39870329-39870329 on Build GRCh38
rs780385460 Chr.19: 39870274-39870275 on Build GRCh38
rs140648438 Chr.19: 39870391-39870391 on Build GRCh38
rs765686847 Chr.19: 39870425-39870425 on Build GRCh38
rs199570976 Chr.19: 39870294-39870294 on Build GRCh38
rs374766909 Chr.19: 39870341-39870341 on Build GRCh38
rs988025387 Chr.19: 39870332-39870332 on Build GRCh38
rs1026537999 Chr.19: 39870299-39870299 on Build GRCh38
rs1053686 Chr.19: 39870288-39870288 on Build GRCh38
rs371263426 Chr.19: 39870319-39870319 on Build GRCh38
rs774614015 Chr.19: 39870442-39870443 on Build GRCh38
rs750965934 Chr.19: 39870342-39870342 on Build GRCh38

View Full Product Details