Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61878068-61878317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS3
Entrez Gene ID
3995
Gene Name
fatty acid desaturase 3
Gene Aliases
CYB5RP, LLCDL3
UniGene
Hs.21765
Gene Chromosome Location
Chr.11: 61873523-61892224 on Build GRCh38
Transcripts
NM_021727.4 XM_011545023.2 XM_017017723.1 XM_017017724.1 NR_106804.1

Gene Symbol
MIR6746
Entrez Gene ID
102465446
Gene Name
microRNA 6746
Gene Aliases
hsa-mir-6746
UniGene
Gene Chromosome Location
Chr.11: 61878216-61878278 on Build GRCh38
Transcripts
NM_021727.4 XM_011545023.2 XM_017017723.1 XM_017017724.1 NR_106804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3450819 c.777C>T p.P259P Chr.11: 61878186-61878186 on Build GRCh38
4897374 c.777C>T p.P259P Chr.11: 61878186-61878186 on Build GRCh38
7280695 c.750T>C p.Y250Y Chr.11: 61878213-61878213 on Build GRCh38
2037985 c.801C>A p.F267L Chr.11: 61878162-61878162 on Build GRCh38
4198675 c.801C>A p.F267L Chr.11: 61878162-61878162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956862983 Chr.11: 61878273-61878273 on Build GRCh38
rs116362087 Chr.11: 61878271-61878271 on Build GRCh38
rs754326244 Chr.11: 61878253-61878254 on Build GRCh38
rs1049288054 Chr.11: 61878099-61878099 on Build GRCh38
rs770308115 Chr.11: 61878199-61878199 on Build GRCh38
rs749566845 Chr.11: 61878150-61878150 on Build GRCh38
rs756176569 Chr.11: 61878257-61878257 on Build GRCh38
rs769363037 Chr.11: 61878284-61878284 on Build GRCh38
rs757053135 Chr.11: 61878243-61878243 on Build GRCh38
rs751394445 Chr.11: 61878238-61878238 on Build GRCh38
rs761666236 Chr.11: 61878296-61878296 on Build GRCh38
rs377717074 Chr.11: 61878229-61878229 on Build GRCh38
rs780254020 Chr.11: 61878148-61878148 on Build GRCh38
rs745928869 Chr.11: 61878144-61878144 on Build GRCh38
rs1001265639 Chr.11: 61878096-61878096 on Build GRCh38
rs773639586 Chr.11: 61878192-61878192 on Build GRCh38
rs370400244 Chr.11: 61878244-61878244 on Build GRCh38
rs751386734 Chr.11: 61878136-61878136 on Build GRCh38
rs776912215 Chr.11: 61878113-61878113 on Build GRCh38
rs750431416 Chr.11: 61878249-61878249 on Build GRCh38
rs370706499 Chr.11: 61878126-61878126 on Build GRCh38
rs764767024 Chr.11: 61878283-61878283 on Build GRCh38
rs759943483 Chr.11: 61878183-61878183 on Build GRCh38
rs766983906 Chr.11: 61878148-61878151 on Build GRCh38
rs183699473 Chr.11: 61878230-61878230 on Build GRCh38
rs772361268 Chr.11: 61878275-61878275 on Build GRCh38
rs549610651 Chr.11: 61878295-61878295 on Build GRCh38
rs531085517 Chr.11: 61878279-61878279 on Build GRCh38
rs764906264 Chr.11: 61878217-61878217 on Build GRCh38
rs762391198 Chr.11: 61878108-61878108 on Build GRCh38
rs763757689 Chr.11: 61878233-61878233 on Build GRCh38
rs759085496 Chr.11: 61878200-61878200 on Build GRCh38
rs376192526 Chr.11: 61878142-61878142 on Build GRCh38
rs140608611 Chr.11: 61878160-61878160 on Build GRCh38
rs756231247 Chr.11: 61878147-61878147 on Build GRCh38
rs1046305809 Chr.11: 61878191-61878191 on Build GRCh38
rs548846159 Chr.11: 61878258-61878258 on Build GRCh38
rs776232730 Chr.11: 61878119-61878119 on Build GRCh38
rs761296196 Chr.11: 61878138-61878139 on Build GRCh38
rs770693836 Chr.11: 61878264-61878266 on Build GRCh38
rs757252065 Chr.11: 61878138-61878138 on Build GRCh38
rs768334070 Chr.11: 61878287-61878287 on Build GRCh38
rs759215085 Chr.11: 61878123-61878123 on Build GRCh38
rs763963678 Chr.11: 61878132-61878132 on Build GRCh38
rs762647475 Chr.11: 61878286-61878286 on Build GRCh38
rs979503654 Chr.11: 61878261-61878261 on Build GRCh38
rs774780631 Chr.11: 61878158-61878158 on Build GRCh38
rs768946836 Chr.11: 61878156-61878156 on Build GRCh38
rs953522166 Chr.11: 61878232-61878232 on Build GRCh38
rs758263162 Chr.11: 61878129-61878129 on Build GRCh38
rs779549009 Chr.11: 61878127-61878127 on Build GRCh38
rs779096471 Chr.11: 61878272-61878272 on Build GRCh38

View Full Product Details