Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 33110555-33110814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LTBP1
Entrez Gene ID
4052
Gene Name
latent transforming growth factor beta binding protein 1
Gene Aliases
-
UniGene
Hs.619315
Gene Chromosome Location
Chr.2: 32947129-33399509 on Build GRCh38
Transcripts
NM_206943.2 XM_005264317.3 XM_005264318.3 XM_011532853.2 XM_011532855.2 XM_011532856.2 XM_011532857.2 XM_011532858.2 XM_011532859.2 XM_011532860.2 XM_011532861.2 XM_011532862.2 XM_017004108.1 XM_017004109.1 XM_017004110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7077743 c.874C>G p.L292V Chr.2: 33110592-33110592 on Build GRCh38
7089887 c.954G>T p.K318N Chr.2: 33110672-33110672 on Build GRCh38
6319399 c.892G>T p.V298L Chr.2: 33110610-33110610 on Build GRCh38
4699436 c.986C>A p.S329Y Chr.2: 33110704-33110704 on Build GRCh38
3581248 c.957G>A p.G319G Chr.2: 33110675-33110675 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911278506 Chr.2: 33110702-33110702 on Build GRCh38
rs761860763 Chr.2: 33110595-33110595 on Build GRCh38
rs774273116 Chr.2: 33110696-33110696 on Build GRCh38
rs557403804 Chr.2: 33110620-33110620 on Build GRCh38
rs564031844 Chr.2: 33110767-33110767 on Build GRCh38
rs748194686 Chr.2: 33110659-33110659 on Build GRCh38
rs546103699 Chr.2: 33110599-33110599 on Build GRCh38
rs767210688 Chr.2: 33110705-33110705 on Build GRCh38
rs752193704 Chr.2: 33110639-33110639 on Build GRCh38
rs759233760 Chr.2: 33110621-33110621 on Build GRCh38
rs780362310 Chr.2: 33110649-33110649 on Build GRCh38
rs752321964 Chr.2: 33110719-33110719 on Build GRCh38
rs747444314 Chr.2: 33110650-33110650 on Build GRCh38
rs992081427 Chr.2: 33110750-33110750 on Build GRCh38
rs747789215 Chr.2: 33110787-33110787 on Build GRCh38
rs17012574 Chr.2: 33110751-33110751 on Build GRCh38
rs117874429 Chr.2: 33110640-33110640 on Build GRCh38
rs540800610 Chr.2: 33110783-33110783 on Build GRCh38
rs775400627 Chr.2: 33110786-33110786 on Build GRCh38
rs766289918 Chr.2: 33110695-33110695 on Build GRCh38
rs955526456 Chr.2: 33110681-33110681 on Build GRCh38
rs218226 Chr.2: 33110780-33110780 on Build GRCh38
rs762962699 Chr.2: 33110693-33110693 on Build GRCh38
rs763486853 Chr.2: 33110732-33110732 on Build GRCh38
rs186071914 Chr.2: 33110763-33110763 on Build GRCh38
rs776691470 Chr.2: 33110594-33110594 on Build GRCh38
rs561396274 Chr.2: 33110744-33110744 on Build GRCh38
rs751450365 Chr.2: 33110610-33110610 on Build GRCh38
rs755344524 Chr.2: 33110743-33110743 on Build GRCh38
rs768856733 Chr.2: 33110653-33110653 on Build GRCh38
rs760180076 Chr.2: 33110725-33110725 on Build GRCh38
rs768903642 Chr.2: 33110592-33110592 on Build GRCh38
rs773155091 Chr.2: 33110668-33110668 on Build GRCh38
rs759417622 Chr.2: 33110704-33110704 on Build GRCh38
rs758950976 Chr.2: 33110645-33110645 on Build GRCh38
rs769796973 Chr.2: 33110662-33110662 on Build GRCh38
rs777685409 Chr.2: 33110760-33110760 on Build GRCh38
rs774432433 Chr.2: 33110778-33110778 on Build GRCh38
rs183318108 Chr.2: 33110738-33110738 on Build GRCh38
rs776459540 Chr.2: 33110770-33110770 on Build GRCh38
rs140100437 Chr.2: 33110641-33110641 on Build GRCh38
rs747205443 Chr.2: 33110586-33110586 on Build GRCh38
rs546203668 Chr.2: 33110752-33110752 on Build GRCh38
rs760409979 Chr.2: 33110757-33110758 on Build GRCh38
rs767335683 Chr.2: 33110629-33110629 on Build GRCh38
rs374136266 Chr.2: 33110737-33110737 on Build GRCh38
rs751765023 Chr.2: 33110732-33110733 on Build GRCh38
rs61751738 Chr.2: 33110742-33110742 on Build GRCh38
rs766301364 Chr.2: 33110609-33110609 on Build GRCh38
rs753426135 Chr.2: 33110733-33110733 on Build GRCh38
rs371301676 Chr.2: 33110601-33110601 on Build GRCh38

View Full Product Details