Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45574781-45575040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH6B
Entrez Gene ID
220081
Gene Name
glutamate rich 6B
Gene Aliases
FAM194B
UniGene
Hs.668747
Gene Chromosome Location
Chr.13: 45541203-45615739 on Build GRCh38
Transcripts
NM_182542.2 XM_011534965.2 XM_017020418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1644589 c.992G>A p.W331* Chr.13: 45574900-45574900 on Build GRCh38
1941620 c.1011G>T p.E337D Chr.13: 45574881-45574881 on Build GRCh38
5790341 c.1050+1G>A p.? Chr.13: 45574841-45574841 on Build GRCh38
6000894 c.1024C>T p.L342L Chr.13: 45574868-45574868 on Build GRCh38
6037673 c.962-5G>A p.? Chr.13: 45574935-45574935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370164131 Chr.13: 45574951-45574951 on Build GRCh38
rs552480295 Chr.13: 45574963-45574963 on Build GRCh38
rs764318385 Chr.13: 45574850-45574850 on Build GRCh38
rs530671784 Chr.13: 45574954-45574954 on Build GRCh38
rs939749628 Chr.13: 45574813-45574814 on Build GRCh38
rs112996787 Chr.13: 45574810-45574811 on Build GRCh38
rs373426079 Chr.13: 45574878-45574878 on Build GRCh38
rs763099557 Chr.13: 45574915-45574915 on Build GRCh38
rs767918255 Chr.13: 45574935-45574935 on Build GRCh38
rs3084021 Chr.13: 45574808-45574810 on Build GRCh38
rs772262728 Chr.13: 45574807-45574808 on Build GRCh38
rs557182075 Chr.13: 45574812-45574812 on Build GRCh38
rs373461118 Chr.13: 45574816-45574816 on Build GRCh38
rs775635999 Chr.13: 45574808-45574808 on Build GRCh38
rs769415587 Chr.13: 45574808-45574809 on Build GRCh38
rs148006228 Chr.13: 45574999-45574999 on Build GRCh38
rs377082518 Chr.13: 45574896-45574896 on Build GRCh38
rs765973169 Chr.13: 45574959-45574959 on Build GRCh38
rs767192895 Chr.13: 45574815-45574816 on Build GRCh38
rs942893 Chr.13: 45574964-45574964 on Build GRCh38
rs535916281 Chr.13: 45574809-45574809 on Build GRCh38
rs879690223 Chr.13: 45574936-45574936 on Build GRCh38
rs564375115 Chr.13: 45574869-45574869 on Build GRCh38
rs752225960 Chr.13: 45574816-45574817 on Build GRCh38
rs922367097 Chr.13: 45574918-45574918 on Build GRCh38
rs547331588 Chr.13: 45575006-45575006 on Build GRCh38
rs984008597 Chr.13: 45574862-45574862 on Build GRCh38
rs1055122875 Chr.13: 45574958-45574958 on Build GRCh38
rs1020018327 Chr.13: 45574861-45574861 on Build GRCh38
rs755806219 Chr.13: 45574819-45574819 on Build GRCh38
rs7982434 Chr.13: 45574953-45574953 on Build GRCh38
rs140782618 Chr.13: 45574809-45574809 on Build GRCh38
rs143466369 Chr.13: 45574854-45574854 on Build GRCh38
rs371645367 Chr.13: 45574832-45574832 on Build GRCh38
rs1046502454 Chr.13: 45574818-45574818 on Build GRCh38
rs373039618 Chr.13: 45574842-45574842 on Build GRCh38
rs953434591 Chr.13: 45574859-45574859 on Build GRCh38
rs770169002 Chr.13: 45574865-45574865 on Build GRCh38
rs543245753 Chr.13: 45574817-45574817 on Build GRCh38
rs191884890 Chr.13: 45574845-45574845 on Build GRCh38
rs576137629 Chr.13: 45574815-45574815 on Build GRCh38
rs201946586 Chr.13: 45574841-45574841 on Build GRCh38
rs568485592 Chr.13: 45574808-45574808 on Build GRCh38
rs754897436 Chr.13: 45574819-45574819 on Build GRCh38
rs952845920 Chr.13: 45574925-45574925 on Build GRCh38
rs774787211 Chr.13: 45574920-45574920 on Build GRCh38
rs200573545 Chr.13: 45574934-45574934 on Build GRCh38
rs1048594723 Chr.13: 45574814-45574814 on Build GRCh38
rs1045804056 Chr.13: 45574986-45574986 on Build GRCh38
rs775826297 Chr.13: 45574810-45574810 on Build GRCh38
rs1010754784 Chr.13: 45574809-45574810 on Build GRCh38
rs759173441 Chr.13: 45574811-45574812 on Build GRCh38
rs985447983 Chr.13: 45574860-45574860 on Build GRCh38
rs185750713 Chr.13: 45574822-45574822 on Build GRCh38
rs181916175 Chr.13: 45574811-45574811 on Build GRCh38
rs972786332 Chr.13: 45574893-45574893 on Build GRCh38

View Full Product Details