Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 157258013-157258238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT5
Entrez Gene ID
11227
Gene Name
polypeptide N-acetylgalactosaminyltransferase 5
Gene Aliases
GALNAC-T5, GALNACT5
UniGene
Hs.269027
Gene Chromosome Location
Chr.2: 157257548-157318491 on Build GRCh38
Transcripts
NM_014568.1 XM_017003237.1 XM_017003238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5642096 c.100C>T p.R34C Chr.2: 157258182-157258182 on Build GRCh38
6271959 c.101G>T p.R34L Chr.2: 157258183-157258183 on Build GRCh38
3364320 c.120C>G p.I40M Chr.2: 157258202-157258202 on Build GRCh38
6669102 c.119T>C p.I40T Chr.2: 157258201-157258201 on Build GRCh38
4582722 c.146T>C p.I49T Chr.2: 157258228-157258228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752478927 Chr.2: 157258189-157258189 on Build GRCh38
rs768981861 Chr.2: 157258170-157258170 on Build GRCh38
rs753401010 Chr.2: 157258214-157258214 on Build GRCh38
rs140376340 Chr.2: 157258183-157258183 on Build GRCh38
rs774703641 Chr.2: 157258061-157258061 on Build GRCh38
rs755844851 Chr.2: 157258200-157258200 on Build GRCh38
rs756713064 Chr.2: 157258122-157258122 on Build GRCh38
rs769453315 Chr.2: 157258036-157258036 on Build GRCh38
rs776714283 Chr.2: 157258171-157258171 on Build GRCh38
rs771407381 Chr.2: 157258055-157258055 on Build GRCh38
rs887724104 Chr.2: 157258166-157258166 on Build GRCh38
rs200904524 Chr.2: 157258109-157258109 on Build GRCh38
rs761786921 Chr.2: 157258096-157258096 on Build GRCh38
rs574772054 Chr.2: 157258075-157258075 on Build GRCh38
rs189039067 Chr.2: 157258065-157258065 on Build GRCh38
rs201844162 Chr.2: 157258099-157258099 on Build GRCh38
rs746351318 Chr.2: 157258139-157258139 on Build GRCh38
rs980718525 Chr.2: 157258069-157258069 on Build GRCh38
rs571258114 Chr.2: 157258120-157258120 on Build GRCh38
rs765213686 Chr.2: 157258181-157258181 on Build GRCh38
rs144724726 Chr.2: 157258136-157258136 on Build GRCh38
rs755722602 Chr.2: 157258119-157258119 on Build GRCh38
rs761997163 Chr.2: 157258173-157258173 on Build GRCh38
rs201732442 Chr.2: 157258076-157258076 on Build GRCh38
rs776715672 Chr.2: 157258095-157258095 on Build GRCh38
rs376299726 Chr.2: 157258090-157258090 on Build GRCh38
rs772446293 Chr.2: 157258140-157258140 on Build GRCh38
rs545992512 Chr.2: 157258108-157258108 on Build GRCh38
rs201125505 Chr.2: 157258182-157258182 on Build GRCh38
rs767421561 Chr.2: 157258186-157258186 on Build GRCh38
rs747247996 Chr.2: 157258160-157258160 on Build GRCh38
rs775790487 Chr.2: 157258150-157258150 on Build GRCh38
rs751515886 Chr.2: 157258103-157258103 on Build GRCh38
rs199520254 Chr.2: 157258044-157258044 on Build GRCh38
rs1037265151 Chr.2: 157258079-157258079 on Build GRCh38
rs928987779 Chr.2: 157258137-157258137 on Build GRCh38
rs754872390 Chr.2: 157258107-157258107 on Build GRCh38
rs760835752 Chr.2: 157258083-157258083 on Build GRCh38
rs749760312 Chr.2: 157258052-157258052 on Build GRCh38
rs763869457 Chr.2: 157258209-157258209 on Build GRCh38
rs757949716 Chr.2: 157258161-157258161 on Build GRCh38

View Full Product Details