Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35444090-35444363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0319L
Entrez Gene ID
79932
Gene Name
KIAA0319 like
Gene Aliases
AAVR
UniGene
Hs.456507
Gene Chromosome Location
Chr.1: 35433490-35557670 on Build GRCh38
Transcripts
NM_024874.4 XM_006710907.1 XM_006710909.1 XM_006710910.1 XM_011542179.2 XM_011542180.1 XM_017002366.1 XM_017002367.1 XM_017002368.1 XM_017002369.1 XM_017002370.1 XM_017002371.1 XM_017002372.1 XM_017002374.1 XM_017002375.1 XM_017002376.1 XM_017002377.1 XM_017002378.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6400728 c.2561T>C p.F854S Chr.1: 35444256-35444256 on Build GRCh38
908692 c.2560T>G p.F854V Chr.1: 35444257-35444257 on Build GRCh38
464498 c.2602C>T p.R868W Chr.1: 35444215-35444215 on Build GRCh38
464499 c.2602C>T p.R868W Chr.1: 35444215-35444215 on Build GRCh38
2080403 c.2545C>T p.P849S Chr.1: 35444272-35444272 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769261314 Chr.1: 35444120-35444121 on Build GRCh38
rs769834330 Chr.1: 35444191-35444191 on Build GRCh38
rs772670478 Chr.1: 35444124-35444125 on Build GRCh38
rs772369259 Chr.1: 35444232-35444232 on Build GRCh38
rs758196396 Chr.1: 35444161-35444161 on Build GRCh38
rs761779434 Chr.1: 35444267-35444267 on Build GRCh38
rs868256159 Chr.1: 35444284-35444284 on Build GRCh38
rs370971124 Chr.1: 35444236-35444236 on Build GRCh38
rs908911921 Chr.1: 35444119-35444119 on Build GRCh38
rs577194387 Chr.1: 35444122-35444122 on Build GRCh38
rs778797057 Chr.1: 35444128-35444128 on Build GRCh38
rs553155302 Chr.1: 35444293-35444293 on Build GRCh38
rs765000757 Chr.1: 35444168-35444168 on Build GRCh38
rs377042074 Chr.1: 35444174-35444174 on Build GRCh38
rs759928836 Chr.1: 35444231-35444231 on Build GRCh38
rs749736814 Chr.1: 35444215-35444215 on Build GRCh38
rs201544013 Chr.1: 35444336-35444336 on Build GRCh38
rs371549694 Chr.1: 35444340-35444340 on Build GRCh38
rs867700756 Chr.1: 35444262-35444262 on Build GRCh38
rs148131828 Chr.1: 35444268-35444268 on Build GRCh38
rs779905420 Chr.1: 35444276-35444276 on Build GRCh38
rs999082576 Chr.1: 35444169-35444169 on Build GRCh38
rs535105734 Chr.1: 35444125-35444125 on Build GRCh38
rs759043835 Chr.1: 35444118-35444118 on Build GRCh38
rs746400668 Chr.1: 35444339-35444339 on Build GRCh38
rs930135268 Chr.1: 35444181-35444181 on Build GRCh38
rs771054832 Chr.1: 35444329-35444329 on Build GRCh38
rs530205030 Chr.1: 35444141-35444141 on Build GRCh38
rs752925371 Chr.1: 35444303-35444303 on Build GRCh38
rs144119734 Chr.1: 35444323-35444323 on Build GRCh38
rs765264102 Chr.1: 35444275-35444275 on Build GRCh38
rs758297310 Chr.1: 35444214-35444214 on Build GRCh38
rs779756181 Chr.1: 35444300-35444300 on Build GRCh38
rs777764304 Chr.1: 35444320-35444320 on Build GRCh38
rs377600647 Chr.1: 35444234-35444234 on Build GRCh38
rs750388192 Chr.1: 35444281-35444281 on Build GRCh38
rs756352014 Chr.1: 35444314-35444314 on Build GRCh38
rs758192624 Chr.1: 35444290-35444290 on Build GRCh38
rs763644672 Chr.1: 35444247-35444247 on Build GRCh38
rs752348415 Chr.1: 35444123-35444123 on Build GRCh38
rs762473435 Chr.1: 35444130-35444131 on Build GRCh38
rs755624576 Chr.1: 35444124-35444124 on Build GRCh38
rs753416410 Chr.1: 35444248-35444248 on Build GRCh38
rs770559112 Chr.1: 35444304-35444304 on Build GRCh38
rs186650518 Chr.1: 35444180-35444180 on Build GRCh38
rs907428814 Chr.1: 35444256-35444256 on Build GRCh38
rs757286382 Chr.1: 35444126-35444126 on Build GRCh38
rs771430922 Chr.1: 35444226-35444226 on Build GRCh38
rs774785464 Chr.1: 35444228-35444228 on Build GRCh38

View Full Product Details