Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27780157-27780386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESCO2
Entrez Gene ID
157570
Gene Name
establishment of sister chromatid cohesion N-acetyltransferase 2
Gene Aliases
2410004I17Rik, EFO2, RBS
UniGene
Hs.99480
Gene Chromosome Location
Chr.8: 27771949-27812404 on Build GRCh38
Transcripts
NM_001017420.2 XM_011544421.2 XM_011544422.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1098787 c.884C>A p.S295Y Chr.8: 27780196-27780196 on Build GRCh38
7232109 c.931G>A p.D311N Chr.8: 27780243-27780243 on Build GRCh38
3432315 c.935C>A p.S312Y Chr.8: 27780247-27780247 on Build GRCh38
4601175 c.923C>T p.S308F Chr.8: 27780235-27780235 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200213698 Chr.8: 27780228-27780228 on Build GRCh38
rs372428854 Chr.8: 27780291-27780291 on Build GRCh38
rs369338792 Chr.8: 27780282-27780282 on Build GRCh38
rs541233922 Chr.8: 27780359-27780360 on Build GRCh38
rs1048556668 Chr.8: 27780201-27780201 on Build GRCh38
rs1034194340 Chr.8: 27780321-27780321 on Build GRCh38
rs780506917 Chr.8: 27780265-27780265 on Build GRCh38
rs890001376 Chr.8: 27780206-27780206 on Build GRCh38
rs777609703 Chr.8: 27780355-27780355 on Build GRCh38
rs760736611 Chr.8: 27780223-27780223 on Build GRCh38
rs80359858 Chr.8: 27780269-27780272 on Build GRCh38
rs797045566 Chr.8: 27780223-27780224 on Build GRCh38
rs760683249 Chr.8: 27780310-27780310 on Build GRCh38
rs1018709387 Chr.8: 27780313-27780313 on Build GRCh38
rs767444182 Chr.8: 27780219-27780219 on Build GRCh38
rs374744436 Chr.8: 27780294-27780294 on Build GRCh38
rs758719753 Chr.8: 27780250-27780250 on Build GRCh38
rs775406849 Chr.8: 27780309-27780309 on Build GRCh38
rs967037326 Chr.8: 27780330-27780330 on Build GRCh38
rs80359857 Chr.8: 27780191-27780192 on Build GRCh38
rs997001624 Chr.8: 27780255-27780255 on Build GRCh38
rs141959652 Chr.8: 27780208-27780208 on Build GRCh38
rs752610227 Chr.8: 27780222-27780222 on Build GRCh38
rs1028476886 Chr.8: 27780281-27780281 on Build GRCh38
rs555057427 Chr.8: 27780271-27780272 on Build GRCh38
rs771735920 Chr.8: 27780301-27780301 on Build GRCh38
rs184200865 Chr.8: 27780227-27780227 on Build GRCh38
rs375459354 Chr.8: 27780248-27780248 on Build GRCh38
rs764127659 Chr.8: 27780318-27780318 on Build GRCh38
rs992776245 Chr.8: 27780337-27780337 on Build GRCh38
rs797045565 Chr.8: 27780206-27780206 on Build GRCh38
rs745639507 Chr.8: 27780297-27780297 on Build GRCh38
rs200848023 Chr.8: 27780215-27780215 on Build GRCh38
rs745837965 Chr.8: 27780297-27780297 on Build GRCh38
rs747449529 Chr.8: 27780267-27780267 on Build GRCh38
rs145734811 Chr.8: 27780241-27780241 on Build GRCh38
rs764991895 Chr.8: 27780243-27780243 on Build GRCh38
rs144259534 Chr.8: 27780337-27780339 on Build GRCh38
rs752508730 Chr.8: 27780194-27780194 on Build GRCh38

View Full Product Details