Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 155929383-155929630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSO
Entrez Gene ID
1519
Gene Name
cathepsin O
Gene Aliases
CTSO1
UniGene
Hs.75262
Gene Chromosome Location
Chr.4: 155924118-155953896 on Build GRCh38
Transcripts
NM_001334.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5038072 c.795A>G p.G265G Chr.4: 155929585-155929585 on Build GRCh38
260327 c.838G>A p.G280R Chr.4: 155929542-155929542 on Build GRCh38
5433337 c.821C>T p.T274I Chr.4: 155929559-155929559 on Build GRCh38
3775653 c.753A>G p.Q251Q Chr.4: 155929627-155929627 on Build GRCh38
5635224 c.796G>T p.E266* Chr.4: 155929584-155929584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765609768 Chr.4: 155929569-155929569 on Build GRCh38
rs958435667 Chr.4: 155929452-155929452 on Build GRCh38
rs199645689 Chr.4: 155929455-155929455 on Build GRCh38
rs751838108 Chr.4: 155929587-155929587 on Build GRCh38
rs758078500 Chr.4: 155929514-155929514 on Build GRCh38
rs753371535 Chr.4: 155929564-155929564 on Build GRCh38
rs995160869 Chr.4: 155929468-155929468 on Build GRCh38
rs1001460715 Chr.4: 155929475-155929475 on Build GRCh38
rs566430890 Chr.4: 155929442-155929442 on Build GRCh38
rs750259093 Chr.4: 155929497-155929497 on Build GRCh38
rs906996063 Chr.4: 155929446-155929446 on Build GRCh38
rs758621406 Chr.4: 155929580-155929580 on Build GRCh38
rs200302394 Chr.4: 155929521-155929521 on Build GRCh38
rs951220766 Chr.4: 155929538-155929538 on Build GRCh38
rs1025422767 Chr.4: 155929534-155929534 on Build GRCh38
rs529076632 Chr.4: 155929436-155929437 on Build GRCh38
rs1055713213 Chr.4: 155929490-155929490 on Build GRCh38
rs1007955798 Chr.4: 155929594-155929594 on Build GRCh38
rs747113984 Chr.4: 155929505-155929505 on Build GRCh38
rs780078489 Chr.4: 155929501-155929501 on Build GRCh38
rs763749060 Chr.4: 155929559-155929559 on Build GRCh38
rs774219606 Chr.4: 155929531-155929531 on Build GRCh38
rs780460172 Chr.4: 155929583-155929583 on Build GRCh38
rs775073825 Chr.4: 155929549-155929549 on Build GRCh38
rs768789376 Chr.4: 155929511-155929511 on Build GRCh38
rs779636509 Chr.4: 155929535-155929535 on Build GRCh38
rs751704284 Chr.4: 155929558-155929558 on Build GRCh38
rs757866604 Chr.4: 155929566-155929566 on Build GRCh38
rs372759572 Chr.4: 155929502-155929502 on Build GRCh38
rs760393290 Chr.4: 155929556-155929556 on Build GRCh38
rs750811108 Chr.4: 155929574-155929574 on Build GRCh38
rs368781823 Chr.4: 155929536-155929536 on Build GRCh38

View Full Product Details