Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64635226-64635499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRXN2
Entrez Gene ID
9379
Gene Name
neurexin 2
Gene Aliases
-
UniGene
Hs.372938
Gene Chromosome Location
Chr.11: 64606174-64723188 on Build GRCh38
Transcripts
NM_015080.3 NM_138732.2 NM_138734.2 XM_005274400.3 XM_005274401.3 XM_005274402.3 XM_011545370.1 XM_011545371.1 XM_011545373.1 XM_011545375.1 XM_011545385.1 XM_017018563.1 XM_017018564.1 XM_017018565.1 XM_017018566.1 XM_017018567.1 XM_017018568.1 XM_017018569.1 XM_017018570.1 XM_017018571.1 XM_017018572.1 XM_017018573.1 XM_017018574.1 XM_017018575.1 XM_017018576.1 XM_017018577.1 XM_017018578.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3666965 c.277T>C p.Y93H Chr.11: 64635441-64635441 on Build GRCh38
6199816 c.3409A>G p.T1137A Chr.11: 64635447-64635447 on Build GRCh38
6730218 c.383G>A p.R128Q Chr.11: 64635335-64635335 on Build GRCh38
4756517 c.3539G>A p.R1180Q Chr.11: 64635317-64635317 on Build GRCh38
139909 c.3425G>A p.G1142E Chr.11: 64635431-64635431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957724593 Chr.11: 64635403-64635403 on Build GRCh38
rs372208408 Chr.11: 64635357-64635357 on Build GRCh38
rs761757989 Chr.11: 64635335-64635335 on Build GRCh38
rs758982262 Chr.11: 64635415-64635415 on Build GRCh38
rs779011613 Chr.11: 64635391-64635391 on Build GRCh38
rs376770323 Chr.11: 64635267-64635267 on Build GRCh38
rs375743651 Chr.11: 64635358-64635358 on Build GRCh38
rs773013797 Chr.11: 64635451-64635451 on Build GRCh38
rs199682650 Chr.11: 64635302-64635302 on Build GRCh38
rs763626243 Chr.11: 64635317-64635317 on Build GRCh38
rs759047742 Chr.11: 64635331-64635331 on Build GRCh38
rs963918721 Chr.11: 64635404-64635404 on Build GRCh38
rs1020873297 Chr.11: 64635425-64635425 on Build GRCh38
rs748621890 Chr.11: 64635259-64635259 on Build GRCh38
rs141426534 Chr.11: 64635426-64635426 on Build GRCh38
rs760480145 Chr.11: 64635456-64635456 on Build GRCh38
rs370108057 Chr.11: 64635392-64635392 on Build GRCh38
rs369081635 Chr.11: 64635346-64635346 on Build GRCh38
rs772355174 Chr.11: 64635261-64635261 on Build GRCh38
rs868859150 Chr.11: 64635366-64635366 on Build GRCh38
rs4290234 Chr.11: 64635341-64635341 on Build GRCh38
rs111679680 Chr.11: 64635421-64635421 on Build GRCh38
rs966912504 Chr.11: 64635436-64635436 on Build GRCh38
rs374697069 Chr.11: 64635309-64635309 on Build GRCh38
rs201851904 Chr.11: 64635459-64635459 on Build GRCh38
rs150390440 Chr.11: 64635446-64635446 on Build GRCh38
rs769021684 Chr.11: 64635301-64635301 on Build GRCh38
rs747464567 Chr.11: 64635417-64635417 on Build GRCh38
rs373059426 Chr.11: 64635450-64635450 on Build GRCh38
rs1026176052 Chr.11: 64635318-64635318 on Build GRCh38
rs199588805 Chr.11: 64635306-64635306 on Build GRCh38
rs767332486 Chr.11: 64635336-64635336 on Build GRCh38
rs761200443 Chr.11: 64635431-64635431 on Build GRCh38
rs996620506 Chr.11: 64635379-64635379 on Build GRCh38
rs373826308 Chr.11: 64635418-64635418 on Build GRCh38
rs755055955 Chr.11: 64635382-64635382 on Build GRCh38
rs771364598 Chr.11: 64635419-64635419 on Build GRCh38
rs148368187 Chr.11: 64635271-64635271 on Build GRCh38

View Full Product Details