Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 60719670-60719937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIX4
Entrez Gene ID
51804
Gene Name
SIX homeobox 4
Gene Aliases
AREC3
UniGene
Hs.690393
Gene Chromosome Location
Chr.14: 60709538-60724321 on Build GRCh38
Transcripts
NM_017420.4 XM_005267759.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5470265 c.1518G>T p.Q506H Chr.14: 60719791-60719791 on Build GRCh38
956751 c.1549G>T p.G517C Chr.14: 60719760-60719760 on Build GRCh38
1370366 c.1506C>A p.F502L Chr.14: 60719803-60719803 on Build GRCh38
2246963 c.1463A>C p.N488T Chr.14: 60719846-60719846 on Build GRCh38
6838621 c.1429C>G p.Q477E Chr.14: 60719880-60719880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776569993 Chr.14: 60719713-60719713 on Build GRCh38
rs761720723 Chr.14: 60719852-60719852 on Build GRCh38
rs754329675 Chr.14: 60719815-60719815 on Build GRCh38
rs984929107 Chr.14: 60719800-60719800 on Build GRCh38
rs1057261805 Chr.14: 60719787-60719787 on Build GRCh38
rs769336965 Chr.14: 60719857-60719857 on Build GRCh38
rs1037884080 Chr.14: 60719719-60719719 on Build GRCh38
rs548235194 Chr.14: 60719753-60719753 on Build GRCh38
rs992150000 Chr.14: 60719698-60719698 on Build GRCh38
rs762565364 Chr.14: 60719755-60719755 on Build GRCh38
rs559052495 Chr.14: 60719705-60719705 on Build GRCh38
rs145386126 Chr.14: 60719861-60719861 on Build GRCh38
rs746131274 Chr.14: 60719830-60719830 on Build GRCh38
rs772416382 Chr.14: 60719834-60719834 on Build GRCh38
rs61743413 Chr.14: 60719841-60719841 on Build GRCh38
rs75128231 Chr.14: 60719758-60719758 on Build GRCh38
rs776459445 Chr.14: 60719847-60719847 on Build GRCh38
rs528198325 Chr.14: 60719715-60719715 on Build GRCh38
rs746012903 Chr.14: 60719818-60719818 on Build GRCh38
rs367915180 Chr.14: 60719795-60719795 on Build GRCh38
rs779112035 Chr.14: 60719827-60719827 on Build GRCh38
rs141516639 Chr.14: 60719784-60719784 on Build GRCh38
rs772707329 Chr.14: 60719865-60719865 on Build GRCh38
rs764682464 Chr.14: 60719749-60719749 on Build GRCh38
rs1000491025 Chr.14: 60719904-60719904 on Build GRCh38
rs370946363 Chr.14: 60719728-60719728 on Build GRCh38
rs1012956373 Chr.14: 60719813-60719813 on Build GRCh38
rs200884734 Chr.14: 60719842-60719842 on Build GRCh38
rs763921345 Chr.14: 60719711-60719711 on Build GRCh38
rs530684279 Chr.14: 60719883-60719883 on Build GRCh38
rs147040477 Chr.14: 60719824-60719824 on Build GRCh38
rs904447933 Chr.14: 60719869-60719869 on Build GRCh38
rs778148156 Chr.14: 60719810-60719810 on Build GRCh38

View Full Product Details