Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144412349-144412574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC39A4
Entrez Gene ID
55630
Gene Name
solute carrier family 39 member 4
Gene Aliases
AEZ, AWMS2, ZIP4
UniGene
Hs.521934
Gene Chromosome Location
Chr.8: 144412414-144416895 on Build GRCh38
Transcripts
NM_001280557.1 NM_017767.2 NM_130849.3 XM_006716599.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3898222 c.1856A>G p.D619G Chr.8: 144412551-144412551 on Build GRCh38
3898221 c.1931A>G p.D644G Chr.8: 144412551-144412551 on Build GRCh38
1265924 c.1835T>C p.L612P Chr.8: 144412572-144412572 on Build GRCh38
1265920 c.1837_1839delCTG p.L613delL Chr.8: 144412568-144412570 on Build GRCh38
1265923 c.1910T>C p.L637P Chr.8: 144412572-144412572 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969827098 Chr.8: 144412421-144412421 on Build GRCh38
rs782794733 Chr.8: 144412498-144412498 on Build GRCh38
rs536723324 Chr.8: 144412471-144412471 on Build GRCh38
rs988769313 Chr.8: 144412479-144412479 on Build GRCh38
rs756243133 Chr.8: 144412454-144412454 on Build GRCh38
rs547713325 Chr.8: 144412375-144412375 on Build GRCh38
rs548280297 Chr.8: 144412431-144412431 on Build GRCh38
rs1023025315 Chr.8: 144412370-144412370 on Build GRCh38
rs200124403 Chr.8: 144412496-144412496 on Build GRCh38
rs978575603 Chr.8: 144412430-144412430 on Build GRCh38
rs781920712 Chr.8: 144412488-144412488 on Build GRCh38
rs369337670 Chr.8: 144412523-144412523 on Build GRCh38
rs200291525 Chr.8: 144412552-144412552 on Build GRCh38
rs200717604 Chr.8: 144412510-144412510 on Build GRCh38
rs529620164 Chr.8: 144412420-144412420 on Build GRCh38
rs782564135 Chr.8: 144412530-144412530 on Build GRCh38
rs1038762995 Chr.8: 144412545-144412545 on Build GRCh38
rs141426507 Chr.8: 144412400-144412400 on Build GRCh38
rs781883858 Chr.8: 144412529-144412529 on Build GRCh38
rs569619977 Chr.8: 144412445-144412445 on Build GRCh38
rs782726592 Chr.8: 144412502-144412502 on Build GRCh38
rs934570845 Chr.8: 144412477-144412477 on Build GRCh38
rs782261765 Chr.8: 144412544-144412544 on Build GRCh38
rs782622118 Chr.8: 144412535-144412535 on Build GRCh38
rs781909019 Chr.8: 144412501-144412501 on Build GRCh38

View Full Product Details