Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47432026-47432299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC8A2
Entrez Gene ID
6543
Gene Name
solute carrier family 8 member A2
Gene Aliases
NCX2
UniGene
Hs.172884
Gene Chromosome Location
Chr.19: 47428017-47472050 on Build GRCh38
Transcripts
NM_015063.2 XM_005259172.1 XM_017027159.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
998733 c.2290G>A p.A764T Chr.19: 47432266-47432266 on Build GRCh38
998732 c.2344T>G p.S782A Chr.19: 47432212-47432212 on Build GRCh38
998731 c.2382C>A p.S794S Chr.19: 47432174-47432174 on Build GRCh38
5856276 c.2371C>G p.L791V Chr.19: 47432185-47432185 on Build GRCh38
6331356 c.2320G>A p.G774S Chr.19: 47432236-47432236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs908810632 Chr.19: 47432249-47432249 on Build GRCh38
rs754179607 Chr.19: 47432183-47432183 on Build GRCh38
rs772891859 Chr.19: 47432279-47432279 on Build GRCh38
rs374763524 Chr.19: 47432121-47432121 on Build GRCh38
rs367751012 Chr.19: 47432264-47432264 on Build GRCh38
rs767573113 Chr.19: 47432146-47432146 on Build GRCh38
rs28637631 Chr.19: 47432222-47432222 on Build GRCh38
rs762817311 Chr.19: 47432131-47432131 on Build GRCh38
rs768429796 Chr.19: 47432118-47432118 on Build GRCh38
rs150808359 Chr.19: 47432267-47432267 on Build GRCh38
rs4801722 Chr.19: 47432095-47432095 on Build GRCh38
rs776476080 Chr.19: 47432266-47432266 on Build GRCh38
rs201973517 Chr.19: 47432255-47432255 on Build GRCh38
rs768924835 Chr.19: 47432067-47432067 on Build GRCh38
rs368246846 Chr.19: 47432128-47432128 on Build GRCh38
rs538645493 Chr.19: 47432113-47432113 on Build GRCh38
rs769578754 Chr.19: 47432278-47432278 on Build GRCh38
rs771570820 Chr.19: 47432096-47432096 on Build GRCh38
rs565171550 Chr.19: 47432093-47432093 on Build GRCh38
rs768413214 Chr.19: 47432259-47432259 on Build GRCh38
rs764356102 Chr.19: 47432177-47432177 on Build GRCh38
rs1053535792 Chr.19: 47432143-47432143 on Build GRCh38
rs760319103 Chr.19: 47432117-47432117 on Build GRCh38
rs746587243 Chr.19: 47432243-47432243 on Build GRCh38
rs867459624 Chr.19: 47432227-47432227 on Build GRCh38
rs779522340 Chr.19: 47432087-47432087 on Build GRCh38
rs771412445 Chr.19: 47432085-47432085 on Build GRCh38
rs778043302 Chr.19: 47432069-47432069 on Build GRCh38
rs781158765 Chr.19: 47432051-47432051 on Build GRCh38
rs758844135 Chr.19: 47432228-47432228 on Build GRCh38
rs757657881 Chr.19: 47432191-47432191 on Build GRCh38
rs752911156 Chr.19: 47432056-47432056 on Build GRCh38
rs779551163 Chr.19: 47432237-47432237 on Build GRCh38
rs200804892 Chr.19: 47432152-47432152 on Build GRCh38
rs756339936 Chr.19: 47432068-47432068 on Build GRCh38
rs779176883 Chr.19: 47432205-47432205 on Build GRCh38
rs375126636 Chr.19: 47432124-47432124 on Build GRCh38
rs532574961 Chr.19: 47432173-47432173 on Build GRCh38
rs374475936 Chr.19: 47432159-47432159 on Build GRCh38
rs895076088 Chr.19: 47432261-47432261 on Build GRCh38

View Full Product Details