Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 101454923-101455150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPC6
Entrez Gene ID
7225
Gene Name
transient receptor potential cation channel subfamily C member 6
Gene Aliases
FSGS2, TRP6
UniGene
Hs.159003
Gene Chromosome Location
Chr.11: 101451470-101584018 on Build GRCh38
Transcripts
NM_004621.5 XM_011542968.2 XM_017018221.1 XM_017018222.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7395222 c.2489G>T p.G830V Chr.11: 101455097-101455097 on Build GRCh38
3808160 c.2485-1G>A p.? Chr.11: 101455102-101455102 on Build GRCh38
3442566 c.2486T>C p.V829A Chr.11: 101455100-101455100 on Build GRCh38
4145010 c.2529C>T p.F843F Chr.11: 101455057-101455057 on Build GRCh38
3383138 c.2524G>A p.D842N Chr.11: 101455062-101455062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377172208 Chr.11: 101455078-101455078 on Build GRCh38
rs370081781 Chr.11: 101455031-101455031 on Build GRCh38
rs149665208 Chr.11: 101455062-101455062 on Build GRCh38
rs570292972 Chr.11: 101455063-101455063 on Build GRCh38
rs754781614 Chr.11: 101455048-101455048 on Build GRCh38
rs766215197 Chr.11: 101455101-101455101 on Build GRCh38
rs746219824 Chr.11: 101455092-101455092 on Build GRCh38
rs772598558 Chr.11: 101454993-101454993 on Build GRCh38
rs756053750 Chr.11: 101455060-101455060 on Build GRCh38
rs201149400 Chr.11: 101455101-101455101 on Build GRCh38
rs917684669 Chr.11: 101454980-101454980 on Build GRCh38
rs878853184 Chr.11: 101455115-101455115 on Build GRCh38
rs779045628 Chr.11: 101455083-101455083 on Build GRCh38
rs751363479 Chr.11: 101455046-101455046 on Build GRCh38
rs772849357 Chr.11: 101455036-101455036 on Build GRCh38
rs61890853 Chr.11: 101455042-101455042 on Build GRCh38
rs757467191 Chr.11: 101455080-101455080 on Build GRCh38
rs746286425 Chr.11: 101454982-101454982 on Build GRCh38
rs202091355 Chr.11: 101455082-101455082 on Build GRCh38
rs145480038 Chr.11: 101455095-101455095 on Build GRCh38
rs774686407 Chr.11: 101454969-101454969 on Build GRCh38
rs950188563 Chr.11: 101454979-101454979 on Build GRCh38
rs747404416 Chr.11: 101455111-101455111 on Build GRCh38
rs199810047 Chr.11: 101455098-101455098 on Build GRCh38
rs367972007 Chr.11: 101455049-101455049 on Build GRCh38
rs199687626 Chr.11: 101455008-101455008 on Build GRCh38
rs762633225 Chr.11: 101455040-101455040 on Build GRCh38
rs72984209 Chr.11: 101455057-101455057 on Build GRCh38
rs949139598 Chr.11: 101454997-101454997 on Build GRCh38
rs761267234 Chr.11: 101455024-101455024 on Build GRCh38

View Full Product Details