Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 55192653-55192898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSL24D1
Entrez Gene ID
51187
Gene Name
ribosomal L24 domain containing 1
Gene Aliases
C15orf15, HRP-L30-iso, L30, RLP24, RPL24, RPL24L, TVAS3
UniGene
Hs.274772
Gene Chromosome Location
Chr.15: 55181314-55197033 on Build GRCh38
Transcripts
NM_016304.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7214833 c.195+8C>T p.? Chr.15: 55192712-55192712 on Build GRCh38
2218217 c.82G>T p.V28L Chr.15: 55192833-55192833 on Build GRCh38
7256329 c.116_117insAA p.N39fs*56 Chr.15: 55192798-55192799 on Build GRCh38
6336859 c.96C>T p.C32C Chr.15: 55192819-55192819 on Build GRCh38
6339525 c.167G>A p.R56Q Chr.15: 55192748-55192748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772358869 Chr.15: 55192700-55192700 on Build GRCh38
rs746665444 Chr.15: 55192707-55192707 on Build GRCh38
rs11540393 Chr.15: 55192786-55192786 on Build GRCh38
rs17851202 Chr.15: 55192827-55192827 on Build GRCh38
rs371969621 Chr.15: 55192862-55192862 on Build GRCh38
rs957387047 Chr.15: 55192828-55192828 on Build GRCh38
rs17852590 Chr.15: 55192782-55192782 on Build GRCh38
rs1020302373 Chr.15: 55192857-55192857 on Build GRCh38
rs561414175 Chr.15: 55192837-55192837 on Build GRCh38
rs779345993 Chr.15: 55192696-55192696 on Build GRCh38
rs886959899 Chr.15: 55192792-55192792 on Build GRCh38
rs766272809 Chr.15: 55192743-55192743 on Build GRCh38
rs11540381 Chr.15: 55192790-55192790 on Build GRCh38
rs78560556 Chr.15: 55192705-55192705 on Build GRCh38
rs534694835 Chr.15: 55192732-55192732 on Build GRCh38
rs1028803526 Chr.15: 55192756-55192756 on Build GRCh38
rs3759863 Chr.15: 55192712-55192712 on Build GRCh38
rs761370511 Chr.15: 55192717-55192717 on Build GRCh38
rs200254994 Chr.15: 55192685-55192685 on Build GRCh38
rs769416449 Chr.15: 55192852-55192852 on Build GRCh38
rs757360650 Chr.15: 55192689-55192689 on Build GRCh38
rs111669877 Chr.15: 55192826-55192826 on Build GRCh38
rs772105282 Chr.15: 55192781-55192781 on Build GRCh38
rs939879035 Chr.15: 55192806-55192806 on Build GRCh38
rs1038188565 Chr.15: 55192846-55192846 on Build GRCh38
rs757258993 Chr.15: 55192801-55192801 on Build GRCh38
rs17856255 Chr.15: 55192830-55192830 on Build GRCh38
rs572620938 Chr.15: 55192784-55192784 on Build GRCh38
rs751351733 Chr.15: 55192770-55192770 on Build GRCh38
rs368427632 Chr.15: 55192711-55192711 on Build GRCh38
rs778975431 Chr.15: 55192805-55192805 on Build GRCh38
rs780645461 Chr.15: 55192840-55192840 on Build GRCh38
rs200023487 Chr.15: 55192832-55192832 on Build GRCh38
rs747817613 Chr.15: 55192847-55192847 on Build GRCh38
rs746390217 Chr.15: 55192697-55192697 on Build GRCh38
rs772741750 Chr.15: 55192786-55192788 on Build GRCh38
rs116077017 Chr.15: 55192723-55192723 on Build GRCh38
rs754119174 Chr.15: 55192785-55192785 on Build GRCh38
rs368924119 Chr.15: 55192740-55192740 on Build GRCh38

View Full Product Details