Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 44732194-44732443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR5
Entrez Gene ID
55615
Gene Name
proline rich 5
Gene Aliases
FLJ20185k, PP610, PROTOR-1, PROTOR1
UniGene
Hs.102336
Gene Chromosome Location
Chr.22: 44668547-44737681 on Build GRCh38
Transcripts
NM_001017528.2 NM_001017529.2 NM_001017530.1 NM_001198721.1 NM_015366.3 NM_181333.3 NM_181334.5

Gene Symbol
PRR5-ARHGAP8
Entrez Gene ID
553158
Gene Name
PRR5-ARHGAP8 readthrough
Gene Aliases
-
UniGene
Hs.102336
Gene Chromosome Location
Chr.22: 44702198-44862784 on Build GRCh38
Transcripts
NM_001017528.2 NM_001017529.2 NM_001017530.1 NM_001198721.1 NM_015366.3 NM_181333.3 NM_181334.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1714468 c.485C>T p.A162V Chr.22: 44732348-44732348 on Build GRCh38
5755772 c.503G>A p.R168H Chr.22: 44732270-44732270 on Build GRCh38
4444870 c.505C>A p.Q169K Chr.22: 44732272-44732272 on Build GRCh38
3062711 c.509A>G p.Q170R Chr.22: 44732372-44732372 on Build GRCh38
3555238 c.504C>T p.I168I Chr.22: 44732367-44732367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764198981 Chr.22: 44732415-44732420 on Build GRCh38
rs777659750 Chr.22: 44732370-44732370 on Build GRCh38
rs763189175 Chr.22: 44732400-44732408 on Build GRCh38
rs765951799 Chr.22: 44732262-44732262 on Build GRCh38
rs926572142 Chr.22: 44732405-44732405 on Build GRCh38
rs142457314 Chr.22: 44732278-44732278 on Build GRCh38
rs778926409 Chr.22: 44732294-44732294 on Build GRCh38
rs754322699 Chr.22: 44732293-44732293 on Build GRCh38
rs763760495 Chr.22: 44732413-44732413 on Build GRCh38
rs771589076 Chr.22: 44732397-44732397 on Build GRCh38
rs773281587 Chr.22: 44732253-44732253 on Build GRCh38
rs756519287 Chr.22: 44732226-44732226 on Build GRCh38
rs572406576 Chr.22: 44732330-44732330 on Build GRCh38
rs751231727 Chr.22: 44732221-44732221 on Build GRCh38
rs765019853 Chr.22: 44732250-44732250 on Build GRCh38
rs201790517 Chr.22: 44732223-44732223 on Build GRCh38
rs765659674 Chr.22: 44732421-44732421 on Build GRCh38
rs370435931 Chr.22: 44732337-44732337 on Build GRCh38
rs759710981 Chr.22: 44732241-44732241 on Build GRCh38
rs375283492 Chr.22: 44732355-44732355 on Build GRCh38
rs754695095 Chr.22: 44732222-44732222 on Build GRCh38
rs746047247 Chr.22: 44732239-44732239 on Build GRCh38
rs375509595 Chr.22: 44732415-44732415 on Build GRCh38
rs5764979 Chr.22: 44732408-44732408 on Build GRCh38
rs757385339 Chr.22: 44732339-44732339 on Build GRCh38
rs774339869 Chr.22: 44732384-44732384 on Build GRCh38
rs758503843 Chr.22: 44732359-44732359 on Build GRCh38
rs750932107 Chr.22: 44732424-44732424 on Build GRCh38
rs778255955 Chr.22: 44732231-44732231 on Build GRCh38
rs8141631 Chr.22: 44732352-44732352 on Build GRCh38
rs750365905 Chr.22: 44732368-44732368 on Build GRCh38
rs769702913 Chr.22: 44732331-44732331 on Build GRCh38
rs752948888 Chr.22: 44732336-44732336 on Build GRCh38
rs1027442030 Chr.22: 44732281-44732281 on Build GRCh38
rs752935264 Chr.22: 44732269-44732269 on Build GRCh38
rs946589794 Chr.22: 44732395-44732395 on Build GRCh38
rs752379834 Chr.22: 44732345-44732345 on Build GRCh38
rs751143808 Chr.22: 44732264-44732264 on Build GRCh38
rs767001335 Chr.22: 44732340-44732340 on Build GRCh38
rs915045412 Chr.22: 44732216-44732216 on Build GRCh38
rs759449236 Chr.22: 44732391-44732391 on Build GRCh38
rs765541168 Chr.22: 44732270-44732271 on Build GRCh38
rs774903791 Chr.22: 44732398-44732398 on Build GRCh38
rs149238292 Chr.22: 44732353-44732353 on Build GRCh38
rs371674855 Chr.22: 44732248-44732248 on Build GRCh38
rs749106643 Chr.22: 44732379-44732379 on Build GRCh38
rs201161761 Chr.22: 44732351-44732351 on Build GRCh38
rs762940353 Chr.22: 44732332-44732332 on Build GRCh38
rs1020993747 Chr.22: 44732228-44732228 on Build GRCh38
rs150104103 Chr.22: 44732338-44732338 on Build GRCh38
rs372358770 Chr.22: 44732414-44732414 on Build GRCh38
rs544764737 Chr.22: 44732301-44732301 on Build GRCh38
rs760293496 Chr.22: 44732350-44732350 on Build GRCh38
rs748564664 Chr.22: 44732224-44732224 on Build GRCh38
rs770923412 Chr.22: 44732383-44732383 on Build GRCh38
rs576042043 Chr.22: 44732226-44732226 on Build GRCh38
rs771214735 Chr.22: 44732222-44732223 on Build GRCh38
rs1055342386 Chr.22: 44732349-44732349 on Build GRCh38
rs763029815 Chr.22: 44732260-44732260 on Build GRCh38
rs555716166 Chr.22: 44732307-44732307 on Build GRCh38
rs776241882 Chr.22: 44732243-44732243 on Build GRCh38
rs756496219 Chr.22: 44732270-44732270 on Build GRCh38
rs148204606 Chr.22: 44732234-44732234 on Build GRCh38
rs563508927 Chr.22: 44732367-44732367 on Build GRCh38
rs749859021 Chr.22: 44732233-44732233 on Build GRCh38
rs199925572 Chr.22: 44732225-44732225 on Build GRCh38
rs956671794 Chr.22: 44732362-44732362 on Build GRCh38
rs772372144 Chr.22: 44732242-44732242 on Build GRCh38
rs137965452 Chr.22: 44732265-44732265 on Build GRCh38
rs761886467 Chr.22: 44732314-44732314 on Build GRCh38

View Full Product Details