Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151874809-151875046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THEM4
Entrez Gene ID
117145
Gene Name
thioesterase superfamily member 4
Gene Aliases
CTMP
UniGene
Hs.164070
Gene Chromosome Location
Chr.1: 151870866-151909885 on Build GRCh38
Transcripts
NM_053055.4 XM_011509142.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
356629 c.693A>G p.I231M Chr.1: 151874918-151874918 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771879954 Chr.1: 151874969-151874971 on Build GRCh38
rs767124334 Chr.1: 151874872-151874872 on Build GRCh38
rs758604035 Chr.1: 151874948-151874948 on Build GRCh38
rs757644614 Chr.1: 151874922-151874922 on Build GRCh38
rs773198128 Chr.1: 151874853-151874853 on Build GRCh38
rs367552448 Chr.1: 151874880-151874880 on Build GRCh38
rs745470903 Chr.1: 151874950-151874950 on Build GRCh38
rs1030861158 Chr.1: 151874986-151874986 on Build GRCh38
rs769538567 Chr.1: 151874845-151874845 on Build GRCh38
rs765878385 Chr.1: 151874938-151874938 on Build GRCh38
rs780414275 Chr.1: 151874956-151874956 on Build GRCh38
rs376827657 Chr.1: 151874858-151874858 on Build GRCh38
rs774588256 Chr.1: 151874991-151874991 on Build GRCh38
rs759075762 Chr.1: 151874863-151874863 on Build GRCh38
rs747938500 Chr.1: 151874838-151874838 on Build GRCh38
rs548305155 Chr.1: 151874962-151874962 on Build GRCh38
rs750503520 Chr.1: 151874943-151874943 on Build GRCh38
rs148860038 Chr.1: 151874857-151874857 on Build GRCh38
rs774125409 Chr.1: 151874931-151874934 on Build GRCh38
rs896177402 Chr.1: 151875012-151875012 on Build GRCh38
rs777375042 Chr.1: 151874965-151874965 on Build GRCh38
rs761083515 Chr.1: 151874884-151874884 on Build GRCh38
rs749131879 Chr.1: 151874966-151874966 on Build GRCh38
rs770235755 Chr.1: 151874871-151874872 on Build GRCh38
rs865919785 Chr.1: 151874978-151874978 on Build GRCh38
rs532959245 Chr.1: 151874959-151874959 on Build GRCh38
rs754231515 Chr.1: 151874892-151874892 on Build GRCh38
rs764429177 Chr.1: 151874887-151874887 on Build GRCh38
rs750898536 Chr.1: 151874861-151874861 on Build GRCh38
rs936394046 Chr.1: 151874878-151874878 on Build GRCh38

View Full Product Details