Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 119936262-119936523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKAP
Entrez Gene ID
79576
Gene Name
NFKB activating protein
Gene Aliases
-
UniGene
Hs.522771
Gene Chromosome Location
Chr.X: 119925050-119943772 on Build GRCh38
Transcripts
NM_024528.3 XM_017029842.1

SNP Detail

SNP ID Chromosome Location
rs759889966 Chr.X: 119936401-119936401 on Build GRCh38
rs769628227 Chr.X: 119936305-119936305 on Build GRCh38
rs150497669 Chr.X: 119936407-119936407 on Build GRCh38
rs374669079 Chr.X: 119936289-119936289 on Build GRCh38
rs747203029 Chr.X: 119936300-119936300 on Build GRCh38
rs190666857 Chr.X: 119936285-119936285 on Build GRCh38
rs754160570 Chr.X: 119936359-119936359 on Build GRCh38
rs194307 Chr.X: 119936448-119936448 on Build GRCh38
rs779818225 Chr.X: 119936306-119936306 on Build GRCh38
rs746456433 Chr.X: 119936476-119936476 on Build GRCh38
rs768490023 Chr.X: 119936318-119936318 on Build GRCh38
rs773044132 Chr.X: 119936334-119936334 on Build GRCh38
rs778751455 Chr.X: 119936458-119936458 on Build GRCh38
rs1013928600 Chr.X: 119936386-119936386 on Build GRCh38
rs765882149 Chr.X: 119936408-119936408 on Build GRCh38
rs10577974 Chr.X: 119936368-119936370 on Build GRCh38
rs761255827 Chr.X: 119936330-119936330 on Build GRCh38
rs371135504 Chr.X: 119936467-119936467 on Build GRCh38
rs570658904 Chr.X: 119936413-119936413 on Build GRCh38
rs761396680 Chr.X: 119936288-119936288 on Build GRCh38
rs749044614 Chr.X: 119936317-119936317 on Build GRCh38
rs748092536 Chr.X: 119936461-119936461 on Build GRCh38
rs112414769 Chr.X: 119936448-119936448 on Build GRCh38
rs373288484 Chr.X: 119936420-119936420 on Build GRCh38
rs866246434 Chr.X: 119936368-119936368 on Build GRCh38
rs779886878 Chr.X: 119936346-119936348 on Build GRCh38
rs765689670 Chr.X: 119936404-119936404 on Build GRCh38
rs945246344 Chr.X: 119936298-119936298 on Build GRCh38
rs754034102 Chr.X: 119936365-119936367 on Build GRCh38
rs73607094 Chr.X: 119936497-119936497 on Build GRCh38
rs774295000 Chr.X: 119936320-119936320 on Build GRCh38
rs202240003 Chr.X: 119936472-119936472 on Build GRCh38
rs771673500 Chr.X: 119936331-119936331 on Build GRCh38
rs757473263 Chr.X: 119936440-119936440 on Build GRCh38
rs760387203 Chr.X: 119936341-119936341 on Build GRCh38
rs964452310 Chr.X: 119936470-119936470 on Build GRCh38
rs764409650 Chr.X: 119936346-119936346 on Build GRCh38
rs750968620 Chr.X: 119936415-119936417 on Build GRCh38
rs369058062 Chr.X: 119936290-119936290 on Build GRCh38
rs754762467 Chr.X: 119936456-119936456 on Build GRCh38

View Full Product Details