Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52502020-52502255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAB1
Entrez Gene ID
23166
Gene Name
stabilin 1
Gene Aliases
CLEVER-1, FEEL-1, FELE-1, FEX1, SCARH2, STAB-1
UniGene
Hs.301989
Gene Chromosome Location
Chr.3: 52495338-52524496 on Build GRCh38
Transcripts
NM_015136.2 XM_005264973.3 XM_005264974.2 XM_006713065.1 XM_017005998.1 XM_017005999.1 XM_017006000.1 XM_017006001.1 XM_017006002.1 XM_017006003.1 XM_017006004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5399754 c.418G>A p.E140K Chr.3: 52502159-52502159 on Build GRCh38
5865011 c.487G>A p.V163M Chr.3: 52502228-52502228 on Build GRCh38
5865010 c.409G>A p.V137M Chr.3: 52502083-52502083 on Build GRCh38
2853715 c.378C>T p.C126C Chr.3: 52502052-52502052 on Build GRCh38
6324476 c.417+10G>A p.? Chr.3: 52502101-52502101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757541695 Chr.3: 52502134-52502134 on Build GRCh38
rs751168593 Chr.3: 52502198-52502198 on Build GRCh38
rs745612141 Chr.3: 52502199-52502199 on Build GRCh38
rs764048212 Chr.3: 52502132-52502132 on Build GRCh38
rs901504030 Chr.3: 52502225-52502225 on Build GRCh38
rs946570167 Chr.3: 52502043-52502043 on Build GRCh38
rs148809253 Chr.3: 52502139-52502139 on Build GRCh38
rs772884237 Chr.3: 52502046-52502046 on Build GRCh38
rs752741111 Chr.3: 52502119-52502119 on Build GRCh38
rs781035831 Chr.3: 52502211-52502211 on Build GRCh38
rs541177060 Chr.3: 52502048-52502048 on Build GRCh38
rs764293623 Chr.3: 52502212-52502212 on Build GRCh38
rs756706101 Chr.3: 52502146-52502146 on Build GRCh38
rs772786796 Chr.3: 52502167-52502167 on Build GRCh38
rs139338725 Chr.3: 52502200-52502200 on Build GRCh38
rs569203616 Chr.3: 52502178-52502178 on Build GRCh38
rs145387328 Chr.3: 52502204-52502204 on Build GRCh38
rs759470226 Chr.3: 52502101-52502101 on Build GRCh38
rs762627573 Chr.3: 52502213-52502213 on Build GRCh38
rs890019652 Chr.3: 52502210-52502210 on Build GRCh38
rs199640409 Chr.3: 52502169-52502169 on Build GRCh38
rs765100827 Chr.3: 52502112-52502112 on Build GRCh38
rs774291979 Chr.3: 52502215-52502215 on Build GRCh38
rs763047511 Chr.3: 52502122-52502122 on Build GRCh38
rs201092493 Chr.3: 52502227-52502227 on Build GRCh38
rs558150044 Chr.3: 52502216-52502216 on Build GRCh38
rs760217624 Chr.3: 52502049-52502049 on Build GRCh38
rs201275556 Chr.3: 52502168-52502168 on Build GRCh38
rs959382728 Chr.3: 52502113-52502113 on Build GRCh38
rs762776022 Chr.3: 52502209-52502209 on Build GRCh38
rs781732531 Chr.3: 52502136-52502136 on Build GRCh38
rs368822018 Chr.3: 52502197-52502197 on Build GRCh38
rs143577711 Chr.3: 52502188-52502188 on Build GRCh38
rs373864526 Chr.3: 52502097-52502097 on Build GRCh38
rs142260689 Chr.3: 52502170-52502170 on Build GRCh38
rs770768274 Chr.3: 52502093-52502093 on Build GRCh38
rs142679283 Chr.3: 52502069-52502069 on Build GRCh38
rs373211349 Chr.3: 52502133-52502133 on Build GRCh38
rs202059385 Chr.3: 52502226-52502226 on Build GRCh38
rs771649805 Chr.3: 52502044-52502044 on Build GRCh38
rs1042574510 Chr.3: 52502063-52502063 on Build GRCh38
rs750035881 Chr.3: 52502149-52502149 on Build GRCh38

View Full Product Details