Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 17590293-17590528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HACD1
Entrez Gene ID
9200
Gene Name
3-hydroxyacyl-CoA dehydratase 1
Gene Aliases
CAP, PTPLA
UniGene
Hs.114062
Gene Chromosome Location
Chr.10: 17589959-17617374 on Build GRCh38
Transcripts
NM_014241.3 XM_005252641.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4013141 c.860A>T p.D287V Chr.10: 17590371-17590371 on Build GRCh38
4444304 c.815G>A p.R272H Chr.10: 17590416-17590416 on Build GRCh38
5430679 c.798C>G p.L266L Chr.10: 17590433-17590433 on Build GRCh38
2131962 c.808A>G p.M270V Chr.10: 17590423-17590423 on Build GRCh38
7191357 c.807T>C p.H269H Chr.10: 17590424-17590424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782343591 Chr.10: 17590436-17590436 on Build GRCh38
rs782722422 Chr.10: 17590478-17590478 on Build GRCh38
rs782008596 Chr.10: 17590485-17590485 on Build GRCh38
rs996079511 Chr.10: 17590395-17590395 on Build GRCh38
rs781989408 Chr.10: 17590426-17590426 on Build GRCh38
rs1018787407 Chr.10: 17590327-17590327 on Build GRCh38
rs782424384 Chr.10: 17590399-17590399 on Build GRCh38
rs782528358 Chr.10: 17590344-17590344 on Build GRCh38
rs368924879 Chr.10: 17590387-17590387 on Build GRCh38
rs565842287 Chr.10: 17590438-17590438 on Build GRCh38
rs781900823 Chr.10: 17590336-17590336 on Build GRCh38
rs59058506 Chr.10: 17590341-17590341 on Build GRCh38
rs143542811 Chr.10: 17590396-17590396 on Build GRCh38
rs149322466 Chr.10: 17590337-17590337 on Build GRCh38
rs541937721 Chr.10: 17590394-17590394 on Build GRCh38
rs781887814 Chr.10: 17590345-17590345 on Build GRCh38
rs983066114 Chr.10: 17590482-17590482 on Build GRCh38
rs1027232748 Chr.10: 17590416-17590416 on Build GRCh38
rs782790178 Chr.10: 17590328-17590328 on Build GRCh38
rs782041782 Chr.10: 17590461-17590461 on Build GRCh38
rs782263866 Chr.10: 17590431-17590431 on Build GRCh38
rs781968830 Chr.10: 17590456-17590456 on Build GRCh38
rs556888412 Chr.10: 17590378-17590378 on Build GRCh38
rs907569739 Chr.10: 17590500-17590500 on Build GRCh38

View Full Product Details