Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 135584697-135584946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP205
Entrez Gene ID
23165
Gene Name
nucleoporin 205
Gene Aliases
C7orf14, NPHS13
UniGene
Hs.743250
Gene Chromosome Location
Chr.7: 135557914-135648753 on Build GRCh38
Transcripts
NM_015135.2 XM_005250235.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3878351 c.1049C>A p.A350E Chr.7: 135584838-135584838 on Build GRCh38
3922765 c.1107C>T p.F369F Chr.7: 135584896-135584896 on Build GRCh38
3878352 c.1101C>T p.N367N Chr.7: 135584890-135584890 on Build GRCh38
75671 c.1121A>T p.E374V Chr.7: 135584910-135584910 on Build GRCh38
71904 c.1043C>G p.A348G Chr.7: 135584832-135584832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776051964 Chr.7: 135584795-135584795 on Build GRCh38
rs746813211 Chr.7: 135584903-135584903 on Build GRCh38
rs750483513 Chr.7: 135584865-135584865 on Build GRCh38
rs550535731 Chr.7: 135584869-135584869 on Build GRCh38
rs906993959 Chr.7: 135584779-135584779 on Build GRCh38
rs757778631 Chr.7: 135584854-135584854 on Build GRCh38
rs548945504 Chr.7: 135584834-135584834 on Build GRCh38
rs547426923 Chr.7: 135584771-135584771 on Build GRCh38
rs371440427 Chr.7: 135584842-135584842 on Build GRCh38
rs759075854 Chr.7: 135584804-135584804 on Build GRCh38
rs567225002 Chr.7: 135584785-135584785 on Build GRCh38
rs968507465 Chr.7: 135584826-135584826 on Build GRCh38
rs772939794 Chr.7: 135584794-135584795 on Build GRCh38
rs995521579 Chr.7: 135584764-135584764 on Build GRCh38
rs780296337 Chr.7: 135584875-135584875 on Build GRCh38
rs374757066 Chr.7: 135584899-135584899 on Build GRCh38
rs139502310 Chr.7: 135584862-135584862 on Build GRCh38
rs1009968333 Chr.7: 135584777-135584777 on Build GRCh38
rs921842418 Chr.7: 135584860-135584860 on Build GRCh38
rs906116054 Chr.7: 135584746-135584746 on Build GRCh38
rs144217755 Chr.7: 135584890-135584890 on Build GRCh38
rs752340377 Chr.7: 135584810-135584810 on Build GRCh38
rs752091216 Chr.7: 135584851-135584851 on Build GRCh38
rs770813366 Chr.7: 135584906-135584906 on Build GRCh38
rs764894904 Chr.7: 135584807-135584807 on Build GRCh38
rs61459701 Chr.7: 135584908-135584908 on Build GRCh38
rs757428765 Chr.7: 135584889-135584889 on Build GRCh38
rs1001603874 Chr.7: 135584752-135584752 on Build GRCh38
rs1031981511 Chr.7: 135584879-135584879 on Build GRCh38
rs760407452 Chr.7: 135584886-135584888 on Build GRCh38
rs1045533788 Chr.7: 135584768-135584768 on Build GRCh38
rs58392569 Chr.7: 135584907-135584907 on Build GRCh38

View Full Product Details