Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 108171251-108171524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR2E1
Entrez Gene ID
7101
Gene Name
nuclear receptor subfamily 2 group E member 1
Gene Aliases
TLL, TLX, XTLL
UniGene
Hs.157688
Gene Chromosome Location
Chr.6: 108166058-108188809 on Build GRCh38
Transcripts
NM_001286102.1 NM_003269.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5474858 c.64C>T p.R22C Chr.6: 108171496-108171496 on Build GRCh38
5908657 c.27C>T p.S9S Chr.6: 108171459-108171459 on Build GRCh38
6729161 c.61G>A p.D21N Chr.6: 108171493-108171493 on Build GRCh38
237858 c.26-8G>A p.? Chr.6: 108171450-108171450 on Build GRCh38
5600070 c.71C>T p.S24L Chr.6: 108171503-108171503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766121249 Chr.6: 108171415-108171415 on Build GRCh38
rs767181240 Chr.6: 108171427-108171427 on Build GRCh38
rs750076132 Chr.6: 108171427-108171438 on Build GRCh38
rs759422145 Chr.6: 108171424-108171424 on Build GRCh38
rs758471698 Chr.6: 108171444-108171444 on Build GRCh38
rs769451088 Chr.6: 108171407-108171407 on Build GRCh38
rs756298274 Chr.6: 108171504-108171504 on Build GRCh38
rs780384211 Chr.6: 108171451-108171451 on Build GRCh38
rs765634214 Chr.6: 108171413-108171414 on Build GRCh38
rs773858504 Chr.6: 108171419-108171419 on Build GRCh38
rs982752438 Chr.6: 108171420-108171420 on Build GRCh38
rs1004692515 Chr.6: 108171304-108171304 on Build GRCh38
rs1018059126 Chr.6: 108171368-108171368 on Build GRCh38
rs750437923 Chr.6: 108171443-108171443 on Build GRCh38
rs753211724 Chr.6: 108171418-108171419 on Build GRCh38
rs201583977 Chr.6: 108171413-108171413 on Build GRCh38
rs545600398 Chr.6: 108171374-108171374 on Build GRCh38
rs752658233 Chr.6: 108171430-108171430 on Build GRCh38
rs142207095 Chr.6: 108171486-108171486 on Build GRCh38
rs2233489 Chr.6: 108171431-108171431 on Build GRCh38
rs915597525 Chr.6: 108171496-108171496 on Build GRCh38
rs188468337 Chr.6: 108171324-108171324 on Build GRCh38
rs1026528118 Chr.6: 108171387-108171387 on Build GRCh38
rs763252794 Chr.6: 108171421-108171428 on Build GRCh38
rs764380616 Chr.6: 108171427-108171427 on Build GRCh38
rs533210216 Chr.6: 108171410-108171410 on Build GRCh38
rs991252299 Chr.6: 108171445-108171445 on Build GRCh38
rs1007622097 Chr.6: 108171290-108171290 on Build GRCh38
rs576491603 Chr.6: 108171291-108171291 on Build GRCh38
rs200851323 Chr.6: 108171426-108171426 on Build GRCh38
rs763744967 Chr.6: 108171275-108171275 on Build GRCh38
rs765317805 Chr.6: 108171440-108171440 on Build GRCh38
rs890513482 Chr.6: 108171280-108171280 on Build GRCh38
rs562555980 Chr.6: 108171385-108171385 on Build GRCh38
rs145563326 Chr.6: 108171402-108171402 on Build GRCh38
rs35730621 Chr.6: 108171367-108171368 on Build GRCh38
rs963393610 Chr.6: 108171326-108171326 on Build GRCh38

View Full Product Details