Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 124371240-124371501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH2D1A
Entrez Gene ID
4068
Gene Name
SH2 domain containing 1A
Gene Aliases
DSHP, EBVS, IMD5, LYP, MTCP1, SAP, SAP/SH2D1A, XLP, XLPD, XLPD1
UniGene
Hs.349094
Gene Chromosome Location
Chr.X: 124346282-124373160 on Build GRCh38
Transcripts
NM_001114937.2 NM_002351.4

SNP Detail

SNP ID Chromosome Location
rs948143513 Chr.X: 124371450-124371450 on Build GRCh38
rs769919066 Chr.X: 124371442-124371443 on Build GRCh38
rs773151816 Chr.X: 124371319-124371320 on Build GRCh38
rs767250216 Chr.X: 124371406-124371406 on Build GRCh38
rs866813603 Chr.X: 124371319-124371319 on Build GRCh38
rs201326965 Chr.X: 124371409-124371409 on Build GRCh38
rs200198093 Chr.X: 124371316-124371320 on Build GRCh38
rs1045022093 Chr.X: 124371456-124371456 on Build GRCh38
rs751985570 Chr.X: 124371293-124371293 on Build GRCh38
rs760630311 Chr.X: 124371311-124371311 on Build GRCh38
rs750980207 Chr.X: 124371419-124371419 on Build GRCh38
rs376444472 Chr.X: 124371317-124371317 on Build GRCh38
rs936697034 Chr.X: 124371472-124371472 on Build GRCh38
rs764986916 Chr.X: 124371339-124371339 on Build GRCh38
rs776527290 Chr.X: 124371312-124371312 on Build GRCh38
rs775191468 Chr.X: 124371386-124371386 on Build GRCh38
rs111033625 Chr.X: 124371389-124371389 on Build GRCh38
rs745892271 Chr.X: 124371405-124371405 on Build GRCh38
rs367998323 Chr.X: 124371395-124371395 on Build GRCh38
rs1042282148 Chr.X: 124371460-124371460 on Build GRCh38
rs763590318 Chr.X: 124371399-124371399 on Build GRCh38
rs991989064 Chr.X: 124371268-124371268 on Build GRCh38

View Full Product Details