Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95736828-95737069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHOBTB3
Entrez Gene ID
22836
Gene Name
Rho related BTB domain containing 3
Gene Aliases
-
UniGene
Hs.445030
Gene Chromosome Location
Chr.5: 95713522-95796367 on Build GRCh38
Transcripts
NM_014899.3 XM_011543279.2 XM_017009236.1 XM_017009237.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1071379 c.237T>C p.F79F Chr.5: 95736897-95736897 on Build GRCh38
6383404 c.392C>A p.A131D Chr.5: 95737052-95737052 on Build GRCh38
3856976 c.276C>T p.G92G Chr.5: 95736936-95736936 on Build GRCh38
6766248 c.285C>A p.I95I Chr.5: 95736945-95736945 on Build GRCh38
5518598 c.303C>T p.N101N Chr.5: 95736963-95736963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768063175 Chr.5: 95737014-95737014 on Build GRCh38
rs762133850 Chr.5: 95737013-95737013 on Build GRCh38
rs199501100 Chr.5: 95736993-95736993 on Build GRCh38
rs751985334 Chr.5: 95737009-95737009 on Build GRCh38
rs765423535 Chr.5: 95736883-95736883 on Build GRCh38
rs779596557 Chr.5: 95736937-95736937 on Build GRCh38
rs766584139 Chr.5: 95736875-95736875 on Build GRCh38
rs759659324 Chr.5: 95736878-95736878 on Build GRCh38
rs755630454 Chr.5: 95736935-95736935 on Build GRCh38
rs752073677 Chr.5: 95736917-95736917 on Build GRCh38
rs749939214 Chr.5: 95736929-95736929 on Build GRCh38
rs773173572 Chr.5: 95736967-95736967 on Build GRCh38
rs758717486 Chr.5: 95736903-95736903 on Build GRCh38
rs1024674800 Chr.5: 95737031-95737031 on Build GRCh38
rs139206452 Chr.5: 95736964-95736964 on Build GRCh38
rs748932209 Chr.5: 95736943-95736943 on Build GRCh38
rs747946829 Chr.5: 95736963-95736963 on Build GRCh38
rs768576337 Chr.5: 95736949-95736949 on Build GRCh38
rs752912885 Chr.5: 95736895-95736895 on Build GRCh38
rs753450088 Chr.5: 95737033-95737033 on Build GRCh38
rs778797272 Chr.5: 95736951-95736951 on Build GRCh38
rs781661099 Chr.5: 95736924-95736924 on Build GRCh38
rs367801787 Chr.5: 95737038-95737038 on Build GRCh38
rs79836860 Chr.5: 95736877-95736877 on Build GRCh38
rs34351922 Chr.5: 95736888-95736888 on Build GRCh38
rs775574971 Chr.5: 95736995-95736995 on Build GRCh38
rs144248494 Chr.5: 95736919-95736919 on Build GRCh38
rs764286650 Chr.5: 95737004-95737004 on Build GRCh38
rs757819060 Chr.5: 95736920-95736920 on Build GRCh38
rs761779560 Chr.5: 95736871-95736871 on Build GRCh38
rs200385981 Chr.5: 95736866-95736866 on Build GRCh38
rs759555357 Chr.5: 95736968-95736968 on Build GRCh38
rs763103033 Chr.5: 95736996-95736996 on Build GRCh38
rs144735826 Chr.5: 95737016-95737016 on Build GRCh38
rs373637979 Chr.5: 95737026-95737026 on Build GRCh38
rs563278102 Chr.5: 95736915-95736915 on Build GRCh38
rs755611194 Chr.5: 95737024-95737024 on Build GRCh38
rs1049450616 Chr.5: 95736956-95736956 on Build GRCh38

View Full Product Details