Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 67009082-67009327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35D1
Entrez Gene ID
23169
Gene Name
solute carrier family 35 member D1
Gene Aliases
UGTREL7
UniGene
Hs.213642
Gene Chromosome Location
Chr.1: 66999332-67054397 on Build GRCh38
Transcripts
NM_015139.2 XM_006710478.1 XM_011541070.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6389766 c.942C>A p.F314L Chr.1: 67009102-67009102 on Build GRCh38
911807 c.904G>A p.V302I Chr.1: 67009140-67009140 on Build GRCh38
5053656 c.930G>A p.T310T Chr.1: 67009114-67009114 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754974284 Chr.1: 67009189-67009189 on Build GRCh38
rs936897641 Chr.1: 67009142-67009142 on Build GRCh38
rs764228619 Chr.1: 67009131-67009131 on Build GRCh38
rs12061148 Chr.1: 67009197-67009197 on Build GRCh38
rs925450718 Chr.1: 67009125-67009125 on Build GRCh38
rs187207257 Chr.1: 67009258-67009258 on Build GRCh38
rs151071375 Chr.1: 67009184-67009184 on Build GRCh38
rs551897062 Chr.1: 67009240-67009240 on Build GRCh38
rs534111616 Chr.1: 67009270-67009270 on Build GRCh38
rs549408335 Chr.1: 67009115-67009115 on Build GRCh38
rs953689589 Chr.1: 67009158-67009158 on Build GRCh38
rs939468014 Chr.1: 67009264-67009264 on Build GRCh38
rs758448658 Chr.1: 67009126-67009126 on Build GRCh38
rs978332731 Chr.1: 67009223-67009223 on Build GRCh38
rs778461128 Chr.1: 67009130-67009132 on Build GRCh38
rs145370126 Chr.1: 67009150-67009150 on Build GRCh38
rs992202216 Chr.1: 67009271-67009274 on Build GRCh38
rs779873232 Chr.1: 67009152-67009152 on Build GRCh38
rs967967459 Chr.1: 67009210-67009210 on Build GRCh38
rs923765361 Chr.1: 67009224-67009224 on Build GRCh38
rs554317550 Chr.1: 67009275-67009275 on Build GRCh38
rs948046528 Chr.1: 67009279-67009279 on Build GRCh38
rs771694261 Chr.1: 67009179-67009184 on Build GRCh38
rs1022207794 Chr.1: 67009199-67009199 on Build GRCh38
rs2755271 Chr.1: 67009294-67009294 on Build GRCh38
rs751766785 Chr.1: 67009140-67009140 on Build GRCh38
rs745564692 Chr.1: 67009152-67009153 on Build GRCh38
rs997951580 Chr.1: 67009267-67009267 on Build GRCh38

View Full Product Details