Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 51808303-51808568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMOD2
Entrez Gene ID
29767
Gene Name
tropomodulin 2
Gene Aliases
N-TMOD, NTMOD
UniGene
Hs.513734
Gene Chromosome Location
Chr.15: 51751561-51816363 on Build GRCh38
Transcripts
NM_001142885.1 NM_014548.3 XM_017022089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1176616 c.1024C>T p.R342C Chr.15: 51808422-51808422 on Build GRCh38
6804484 c.1025G>A p.R342H Chr.15: 51808423-51808423 on Build GRCh38
1373455 c.1048C>T p.R350* Chr.15: 51808446-51808446 on Build GRCh38
4881361 c.1053G>C p.R351S Chr.15: 51808451-51808451 on Build GRCh38
5342583 c.1049G>A p.R350Q Chr.15: 51808447-51808447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553911663 Chr.15: 51808539-51808539 on Build GRCh38
rs771097615 Chr.15: 51808471-51808471 on Build GRCh38
rs775111938 Chr.15: 51808417-51808417 on Build GRCh38
rs1006938590 Chr.15: 51808457-51808457 on Build GRCh38
rs372135721 Chr.15: 51808376-51808376 on Build GRCh38
rs749479890 Chr.15: 51808381-51808381 on Build GRCh38
rs139977930 Chr.15: 51808407-51808407 on Build GRCh38
rs758385185 Chr.15: 51808444-51808444 on Build GRCh38
rs779836040 Chr.15: 51808446-51808446 on Build GRCh38
rs916252344 Chr.15: 51808344-51808344 on Build GRCh38
rs756513904 Chr.15: 51808354-51808354 on Build GRCh38
rs760070721 Chr.15: 51808422-51808422 on Build GRCh38
rs1051720958 Chr.15: 51808442-51808442 on Build GRCh38
rs774627169 Chr.15: 51808405-51808405 on Build GRCh38
rs371289123 Chr.15: 51808447-51808447 on Build GRCh38
rs755344306 Chr.15: 51808403-51808404 on Build GRCh38
rs749724202 Chr.15: 51808387-51808387 on Build GRCh38
rs772672117 Chr.15: 51808478-51808478 on Build GRCh38
rs772407580 Chr.15: 51808411-51808411 on Build GRCh38
rs775855006 Chr.15: 51808493-51808493 on Build GRCh38
rs1018193710 Chr.15: 51808505-51808505 on Build GRCh38
rs778031233 Chr.15: 51808460-51808460 on Build GRCh38
rs749530140 Chr.15: 51808466-51808466 on Build GRCh38
rs552844883 Chr.15: 51808519-51808519 on Build GRCh38
rs781663661 Chr.15: 51808463-51808465 on Build GRCh38
rs754692450 Chr.15: 51808379-51808379 on Build GRCh38
rs756147137 Chr.15: 51808452-51808452 on Build GRCh38
rs777892061 Chr.15: 51808375-51808375 on Build GRCh38
rs750205036 Chr.15: 51808441-51808441 on Build GRCh38
rs765129073 Chr.15: 51808439-51808439 on Build GRCh38
rs753526892 Chr.15: 51808429-51808429 on Build GRCh38
rs760290202 Chr.15: 51808504-51808504 on Build GRCh38
rs771277142 Chr.15: 51808393-51808393 on Build GRCh38
rs763904794 Chr.15: 51808423-51808423 on Build GRCh38
rs761499393 Chr.15: 51808432-51808432 on Build GRCh38
rs746192601 Chr.15: 51808475-51808475 on Build GRCh38

View Full Product Details