Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160171210-160171459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A4
Entrez Gene ID
480
Gene Name
ATPase Na+/K+ transporting subunit alpha 4
Gene Aliases
ATP1A1, ATP1AL2
UniGene
Hs.662219
Gene Chromosome Location
Chr.1: 160151562-160186977 on Build GRCh38
Transcripts
NM_144699.3 XM_011509582.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
339379 c.1607C>G p.S536* Chr.1: 160171366-160171366 on Build GRCh38
4470030 c.1635C>T p.F545F Chr.1: 160171394-160171394 on Build GRCh38
5733986 c.1508G>A p.R503Q Chr.1: 160171267-160171267 on Build GRCh38
1688950 c.1615G>A p.D539N Chr.1: 160171374-160171374 on Build GRCh38
337617 c.1500C>T p.I500I Chr.1: 160171259-160171259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200739116 Chr.1: 160171333-160171333 on Build GRCh38
rs760910264 Chr.1: 160171274-160171274 on Build GRCh38
rs762637659 Chr.1: 160171431-160171431 on Build GRCh38
rs761721153 Chr.1: 160171289-160171289 on Build GRCh38
rs755731268 Chr.1: 160171424-160171424 on Build GRCh38
rs150252500 Chr.1: 160171372-160171372 on Build GRCh38
rs776243597 Chr.1: 160171284-160171284 on Build GRCh38
rs113494317 Chr.1: 160171363-160171363 on Build GRCh38
rs16831482 Chr.1: 160171381-160171381 on Build GRCh38
rs143272416 Chr.1: 160171279-160171279 on Build GRCh38
rs772708143 Chr.1: 160171364-160171364 on Build GRCh38
rs765819014 Chr.1: 160171304-160171304 on Build GRCh38
rs113220471 Chr.1: 160171323-160171323 on Build GRCh38
rs768982750 Chr.1: 160171373-160171373 on Build GRCh38
rs775722693 Chr.1: 160171269-160171269 on Build GRCh38
rs771320642 Chr.1: 160171240-160171240 on Build GRCh38
rs772331708 Chr.1: 160171267-160171267 on Build GRCh38
rs774962487 Chr.1: 160171260-160171260 on Build GRCh38
rs977494202 Chr.1: 160171416-160171416 on Build GRCh38
rs752449048 Chr.1: 160171420-160171420 on Build GRCh38
rs142215427 Chr.1: 160171313-160171313 on Build GRCh38
rs868727855 Chr.1: 160171438-160171438 on Build GRCh38
rs774564912 Chr.1: 160171266-160171266 on Build GRCh38
rs137950481 Chr.1: 160171405-160171405 on Build GRCh38
rs625549 Chr.1: 160171406-160171406 on Build GRCh38
rs886855479 Chr.1: 160171349-160171349 on Build GRCh38
rs780944584 Chr.1: 160171316-160171316 on Build GRCh38
rs148327673 Chr.1: 160171280-160171280 on Build GRCh38
rs746317383 Chr.1: 160171356-160171356 on Build GRCh38
rs1004000515 Chr.1: 160171366-160171366 on Build GRCh38
rs919475096 Chr.1: 160171265-160171265 on Build GRCh38
rs779267926 Chr.1: 160171425-160171425 on Build GRCh38
rs764444541 Chr.1: 160171281-160171281 on Build GRCh38
rs751074976 Chr.1: 160171312-160171312 on Build GRCh38
rs750442801 Chr.1: 160171290-160171290 on Build GRCh38
rs973557553 Chr.1: 160171376-160171376 on Build GRCh38
rs750272358 Chr.1: 160171424-160171424 on Build GRCh38
rs758928463 Chr.1: 160171437-160171437 on Build GRCh38
rs147697970 Chr.1: 160171371-160171371 on Build GRCh38
rs776955295 Chr.1: 160171375-160171375 on Build GRCh38
rs935822119 Chr.1: 160171432-160171432 on Build GRCh38
rs759087766 Chr.1: 160171417-160171417 on Build GRCh38
rs138454276 Chr.1: 160171403-160171403 on Build GRCh38
rs757890752 Chr.1: 160171300-160171300 on Build GRCh38
rs779270944 Chr.1: 160171344-160171344 on Build GRCh38
rs528042190 Chr.1: 160171413-160171413 on Build GRCh38
rs946193022 Chr.1: 160171233-160171233 on Build GRCh38
rs769689108 Chr.1: 160171390-160171390 on Build GRCh38
rs767071083 Chr.1: 160171419-160171419 on Build GRCh38

View Full Product Details