Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122204532-122204771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT4
Entrez Gene ID
79369
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4
Gene Aliases
B3GN-T4, beta3Gn-T4
UniGene
Hs.363315
Gene Chromosome Location
Chr.12: 122203681-122208135 on Build GRCh38
Transcripts
NM_030765.2 XM_011538732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4402436 c.5T>G p.L2R Chr.12: 122204623-122204623 on Build GRCh38
3792281 c.19T>G p.S7A Chr.12: 122204637-122204637 on Build GRCh38
3457292 c.55C>T p.L19F Chr.12: 122204673-122204673 on Build GRCh38
4997997 c.16C>G p.P6A Chr.12: 122204634-122204634 on Build GRCh38
4664884 c.52G>T p.G18C Chr.12: 122204670-122204670 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966936863 Chr.12: 122204561-122204561 on Build GRCh38
rs762542367 Chr.12: 122204623-122204623 on Build GRCh38
rs764706708 Chr.12: 122204580-122204580 on Build GRCh38
rs544378961 Chr.12: 122204604-122204604 on Build GRCh38
rs777131312 Chr.12: 122204703-122204703 on Build GRCh38
rs750960027 Chr.12: 122204574-122204574 on Build GRCh38
rs757076167 Chr.12: 122204593-122204593 on Build GRCh38
rs779021474 Chr.12: 122204728-122204728 on Build GRCh38
rs201631024 Chr.12: 122204732-122204732 on Build GRCh38
rs757733467 Chr.12: 122204718-122204718 on Build GRCh38
rs762326610 Chr.12: 122204712-122204712 on Build GRCh38
rs368808281 Chr.12: 122204597-122204597 on Build GRCh38
rs145969982 Chr.12: 122204615-122204615 on Build GRCh38
rs185977606 Chr.12: 122204700-122204700 on Build GRCh38
rs994191249 Chr.12: 122204566-122204566 on Build GRCh38
rs751253220 Chr.12: 122204630-122204630 on Build GRCh38
rs761148838 Chr.12: 122204579-122204580 on Build GRCh38
rs12828049 Chr.12: 122204697-122204697 on Build GRCh38
rs767846047 Chr.12: 122204573-122204573 on Build GRCh38
rs371092163 Chr.12: 122204752-122204752 on Build GRCh38
rs981718654 Chr.12: 122204750-122204750 on Build GRCh38
rs748382395 Chr.12: 122204691-122204691 on Build GRCh38
rs550784704 Chr.12: 122204722-122204722 on Build GRCh38
rs1013180861 Chr.12: 122204727-122204727 on Build GRCh38
rs1049444784 Chr.12: 122204649-122204649 on Build GRCh38
rs751258059 Chr.12: 122204585-122204585 on Build GRCh38
rs780248621 Chr.12: 122204600-122204600 on Build GRCh38
rs781232010 Chr.12: 122204595-122204595 on Build GRCh38
rs763590068 Chr.12: 122204625-122204625 on Build GRCh38
rs1047930584 Chr.12: 122204710-122204710 on Build GRCh38
rs750280847 Chr.12: 122204650-122204650 on Build GRCh38
rs12827843 Chr.12: 122204614-122204614 on Build GRCh38
rs377471867 Chr.12: 122204588-122204588 on Build GRCh38
rs374352251 Chr.12: 122204687-122204687 on Build GRCh38
rs763678288 Chr.12: 122204713-122204713 on Build GRCh38
rs768747434 Chr.12: 122204605-122204605 on Build GRCh38
rs764535548 Chr.12: 122204592-122204593 on Build GRCh38
rs761492332 Chr.12: 122204714-122204714 on Build GRCh38
rs774645996 Chr.12: 122204606-122204606 on Build GRCh38
rs375650657 Chr.12: 122204735-122204735 on Build GRCh38
rs374677108 Chr.12: 122204562-122204563 on Build GRCh38
rs971365477 Chr.12: 122204744-122204744 on Build GRCh38
rs756949895 Chr.12: 122204632-122204632 on Build GRCh38
rs747434133 Chr.12: 122204698-122204698 on Build GRCh38
rs202226254 Chr.12: 122204584-122204584 on Build GRCh38
rs201510031 Chr.12: 122204607-122204607 on Build GRCh38
rs757985534 Chr.12: 122204583-122204583 on Build GRCh38
rs200385749 Chr.12: 122204675-122204675 on Build GRCh38
rs199704684 Chr.12: 122204715-122204715 on Build GRCh38
rs749268524 Chr.12: 122204672-122204672 on Build GRCh38
rs925992856 Chr.12: 122204570-122204570 on Build GRCh38
rs756115422 Chr.12: 122204652-122204652 on Build GRCh38
rs772321910 Chr.12: 122204693-122204693 on Build GRCh38
rs372784600 Chr.12: 122204562-122204562 on Build GRCh38
rs752192275 Chr.12: 122204621-122204621 on Build GRCh38
rs7136356 Chr.12: 122204634-122204634 on Build GRCh38
rs780196783 Chr.12: 122204660-122204660 on Build GRCh38
rs367733493 Chr.12: 122204581-122204581 on Build GRCh38

View Full Product Details