Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70832847-70833108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD207
Entrez Gene ID
50489
Gene Name
CD207 molecule
Gene Aliases
CLEC4K
UniGene
Hs.199731
Gene Chromosome Location
Chr.2: 70825248-70860787 on Build GRCh38
Transcripts
NM_015717.4 XM_011532874.1 XM_011532875.2 XM_011532876.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3724397 c.613_614insG p.N205fs*14 Chr.2: 70833003-70833004 on Build GRCh38
3739896 c.566-6C>T p.? Chr.2: 70833057-70833057 on Build GRCh38
1022550 c.649T>C p.Y217H Chr.2: 70832968-70832968 on Build GRCh38
4095383 c.630T>C p.S210S Chr.2: 70832987-70832987 on Build GRCh38
6542696 c.674C>A p.S225Y Chr.2: 70832943-70832943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542384792 Chr.2: 70832876-70832876 on Build GRCh38
rs559383492 Chr.2: 70832879-70832879 on Build GRCh38
rs781970590 Chr.2: 70832922-70832922 on Build GRCh38
rs782816989 Chr.2: 70832969-70832969 on Build GRCh38
rs782225816 Chr.2: 70832976-70832976 on Build GRCh38
rs1041806148 Chr.2: 70833071-70833071 on Build GRCh38
rs782137807 Chr.2: 70832884-70832884 on Build GRCh38
rs782617460 Chr.2: 70832975-70832975 on Build GRCh38
rs781901575 Chr.2: 70832890-70832890 on Build GRCh38
rs782515098 Chr.2: 70832978-70832978 on Build GRCh38
rs782391351 Chr.2: 70832926-70832926 on Build GRCh38
rs6712863 Chr.2: 70832906-70832906 on Build GRCh38
rs17006436 Chr.2: 70832980-70832980 on Build GRCh38
rs782627815 Chr.2: 70832904-70832904 on Build GRCh38
rs781862128 Chr.2: 70832880-70832880 on Build GRCh38
rs150805721 Chr.2: 70833078-70833078 on Build GRCh38
rs782694463 Chr.2: 70833072-70833072 on Build GRCh38
rs781942625 Chr.2: 70832869-70832869 on Build GRCh38
rs901383724 Chr.2: 70833063-70833063 on Build GRCh38
rs781788022 Chr.2: 70833077-70833077 on Build GRCh38
rs782247148 Chr.2: 70832925-70832925 on Build GRCh38
rs781919980 Chr.2: 70832992-70832992 on Build GRCh38
rs782565816 Chr.2: 70832920-70832920 on Build GRCh38
rs782219999 Chr.2: 70833081-70833081 on Build GRCh38
rs781829207 Chr.2: 70833044-70833044 on Build GRCh38
rs573751884 Chr.2: 70832866-70832866 on Build GRCh38
rs782015589 Chr.2: 70833034-70833034 on Build GRCh38
rs782717829 Chr.2: 70833041-70833041 on Build GRCh38
rs376010760 Chr.2: 70832959-70832959 on Build GRCh38
rs782417032 Chr.2: 70832985-70832985 on Build GRCh38
rs782230504 Chr.2: 70833080-70833080 on Build GRCh38
rs782427845 Chr.2: 70833014-70833014 on Build GRCh38
rs199605575 Chr.2: 70833026-70833026 on Build GRCh38
rs563353094 Chr.2: 70832865-70832865 on Build GRCh38
rs781850322 Chr.2: 70832960-70832960 on Build GRCh38
rs782531490 Chr.2: 70833065-70833065 on Build GRCh38
rs533675173 Chr.2: 70832972-70832972 on Build GRCh38
rs374676643 Chr.2: 70832918-70832918 on Build GRCh38
rs1041278596 Chr.2: 70833061-70833061 on Build GRCh38
rs782792485 Chr.2: 70832885-70832885 on Build GRCh38
rs373032299 Chr.2: 70833059-70833059 on Build GRCh38
rs782020363 Chr.2: 70832936-70832936 on Build GRCh38
rs781873105 Chr.2: 70833062-70833062 on Build GRCh38
rs782697700 Chr.2: 70832939-70832939 on Build GRCh38
rs782163497 Chr.2: 70832937-70832937 on Build GRCh38
rs782056019 Chr.2: 70833038-70833038 on Build GRCh38
rs782482178 Chr.2: 70832901-70832901 on Build GRCh38
rs372167399 Chr.2: 70832945-70832945 on Build GRCh38
rs367897796 Chr.2: 70832899-70832899 on Build GRCh38
rs782643609 Chr.2: 70832979-70832979 on Build GRCh38
rs782104857 Chr.2: 70833015-70833015 on Build GRCh38

View Full Product Details