Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43432818-43433049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC10
Entrez Gene ID
89845
Gene Name
ATP binding cassette subfamily C member 10
Gene Aliases
EST182763, MRP7, SIMRP7
UniGene
Hs.55879
Gene Chromosome Location
Chr.6: 43427538-43451994 on Build GRCh38
Transcripts
NM_001198934.1 NM_033450.2 XM_011514974.2 XM_011514983.2 XM_017011445.1 XM_017011446.1 XM_017011447.1 XM_017011448.1 XM_017011449.1 XM_017011450.1 XM_017011451.1 XM_017011452.1 XM_017011453.1 XM_017011454.1 XM_017011455.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6197799 c.1039G>T p.E347* Chr.6: 43433019-43433019 on Build GRCh38
6529806 c.1029G>C p.Q343H Chr.6: 43433009-43433009 on Build GRCh38
298887 c.865G>A p.G289S Chr.6: 43432974-43432974 on Build GRCh38
6174554 c.948G>C p.G316G Chr.6: 43432928-43432928 on Build GRCh38
5648794 c.990A>T p.L330L Chr.6: 43432970-43432970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756843484 Chr.6: 43432870-43432870 on Build GRCh38
rs976949093 Chr.6: 43432964-43432964 on Build GRCh38
rs559072013 Chr.6: 43432984-43432984 on Build GRCh38
rs145139535 Chr.6: 43433015-43433015 on Build GRCh38
rs768546118 Chr.6: 43432993-43432993 on Build GRCh38
rs762877448 Chr.6: 43432930-43432930 on Build GRCh38
rs754124384 Chr.6: 43432872-43432872 on Build GRCh38
rs758767831 Chr.6: 43432897-43432897 on Build GRCh38
rs755538846 Chr.6: 43432888-43432888 on Build GRCh38
rs767100799 Chr.6: 43432872-43432872 on Build GRCh38
rs202203665 Chr.6: 43432860-43432860 on Build GRCh38
rs776805314 Chr.6: 43432918-43432918 on Build GRCh38
rs145760020 Chr.6: 43432946-43432946 on Build GRCh38
rs547806531 Chr.6: 43432882-43432882 on Build GRCh38
rs538867404 Chr.6: 43432962-43432962 on Build GRCh38
rs759593725 Chr.6: 43432968-43432968 on Build GRCh38
rs200276315 Chr.6: 43432937-43432937 on Build GRCh38
rs745888504 Chr.6: 43432924-43432924 on Build GRCh38
rs781216160 Chr.6: 43432973-43432973 on Build GRCh38
rs150374334 Chr.6: 43433005-43433005 on Build GRCh38
rs751283694 Chr.6: 43432866-43432866 on Build GRCh38
rs1012826581 Chr.6: 43432887-43432887 on Build GRCh38
rs868688148 Chr.6: 43432868-43432868 on Build GRCh38
rs201484865 Chr.6: 43432844-43432844 on Build GRCh38
rs745967911 Chr.6: 43432974-43432974 on Build GRCh38
rs749268989 Chr.6: 43432991-43432991 on Build GRCh38
rs1018318507 Chr.6: 43432902-43432902 on Build GRCh38
rs748535408 Chr.6: 43432891-43432891 on Build GRCh38
rs765048084 Chr.6: 43432853-43432853 on Build GRCh38
rs769786378 Chr.6: 43432928-43432928 on Build GRCh38
rs952403231 Chr.6: 43432920-43432920 on Build GRCh38
rs775342917 Chr.6: 43432929-43432929 on Build GRCh38
rs140348023 Chr.6: 43432947-43432947 on Build GRCh38
rs555034468 Chr.6: 43433022-43433022 on Build GRCh38
rs760370204 Chr.6: 43433027-43433027 on Build GRCh38
rs763744091 Chr.6: 43432865-43432865 on Build GRCh38
rs758960596 Chr.6: 43432955-43432955 on Build GRCh38
rs375765975 Chr.6: 43432979-43432979 on Build GRCh38
rs771028569 Chr.6: 43432913-43432913 on Build GRCh38
rs765842062 Chr.6: 43432950-43432950 on Build GRCh38
rs371159732 Chr.6: 43432901-43432901 on Build GRCh38
rs775287791 Chr.6: 43432855-43432855 on Build GRCh38
rs146848280 Chr.6: 43433025-43433025 on Build GRCh38
rs147818692 Chr.6: 43432898-43432898 on Build GRCh38
rs760044039 Chr.6: 43432949-43432949 on Build GRCh38
rs779511674 Chr.6: 43432889-43432889 on Build GRCh38
rs751830941 Chr.6: 43432957-43432957 on Build GRCh38
rs768502452 Chr.6: 43432932-43432932 on Build GRCh38
rs73424611 Chr.6: 43432953-43432953 on Build GRCh38
rs144342445 Chr.6: 43432896-43432896 on Build GRCh38

View Full Product Details