Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81240862-81241121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf89
Entrez Gene ID
284184
Gene Name
chromosome 17 open reading frame 89
Gene Aliases
-
UniGene
Hs.356545
Gene Chromosome Location
Chr.17: 81239311-81241298 on Build GRCh38
Transcripts
NM_001086521.1

SNP Detail

SNP ID Chromosome Location
rs942667899 Chr.17: 81240950-81240950 on Build GRCh38
rs1002804031 Chr.17: 81241008-81241008 on Build GRCh38
rs371310918 Chr.17: 81240994-81240994 on Build GRCh38
rs368335720 Chr.17: 81240910-81240910 on Build GRCh38
rs912574448 Chr.17: 81240924-81240924 on Build GRCh38
rs375816631 Chr.17: 81240938-81240938 on Build GRCh38
rs113490148 Chr.17: 81241094-81241094 on Build GRCh38
rs878899849 Chr.17: 81240909-81240909 on Build GRCh38
rs369042116 Chr.17: 81241088-81241088 on Build GRCh38
rs757341965 Chr.17: 81240966-81240966 on Build GRCh38
rs370128387 Chr.17: 81240974-81240974 on Build GRCh38
rs765130289 Chr.17: 81240972-81240972 on Build GRCh38
rs955219872 Chr.17: 81240905-81240905 on Build GRCh38
rs777099199 Chr.17: 81241021-81241021 on Build GRCh38
rs930208096 Chr.17: 81240913-81240913 on Build GRCh38
rs747659979 Chr.17: 81241000-81241000 on Build GRCh38
rs373091024 Chr.17: 81240937-81240937 on Build GRCh38
rs537110946 Chr.17: 81240911-81240911 on Build GRCh38
rs753926266 Chr.17: 81240946-81240946 on Build GRCh38
rs59124084 Chr.17: 81240922-81240922 on Build GRCh38
rs539090122 Chr.17: 81241095-81241095 on Build GRCh38
rs764554149 Chr.17: 81241073-81241073 on Build GRCh38
rs779834662 Chr.17: 81240988-81240988 on Build GRCh38
rs958364165 Chr.17: 81241013-81241013 on Build GRCh38
rs1016463270 Chr.17: 81241038-81241038 on Build GRCh38
rs1035574018 Chr.17: 81241009-81241009 on Build GRCh38
rs555208393 Chr.17: 81241090-81241090 on Build GRCh38
rs764409687 Chr.17: 81240940-81240940 on Build GRCh38
rs761897458 Chr.17: 81241083-81241083 on Build GRCh38
rs375853381 Chr.17: 81240998-81240998 on Build GRCh38
rs776067909 Chr.17: 81241051-81241051 on Build GRCh38
rs893216444 Chr.17: 81240975-81240976 on Build GRCh38
rs746401977 Chr.17: 81241030-81241030 on Build GRCh38
rs957182784 Chr.17: 81240917-81240917 on Build GRCh38
rs756694869 Chr.17: 81241069-81241069 on Build GRCh38
rs1018433859 Chr.17: 81240912-81240912 on Build GRCh38
rs570351619 Chr.17: 81240997-81240997 on Build GRCh38
rs997739822 Chr.17: 81240887-81240887 on Build GRCh38
rs557651590 Chr.17: 81240897-81240897 on Build GRCh38
rs769378487 Chr.17: 81241019-81241019 on Build GRCh38
rs767994583 Chr.17: 81240973-81240973 on Build GRCh38
rs754302658 Chr.17: 81241099-81241099 on Build GRCh38
rs772642620 Chr.17: 81241040-81241040 on Build GRCh38
rs776885480 Chr.17: 81241080-81241080 on Build GRCh38

View Full Product Details