Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35059015-35059272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VCP
Entrez Gene ID
7415
Gene Name
valosin containing protein
Gene Aliases
ALS14, CMT2Y, HEL-220, HEL-S-70, IBMPFD, IBMPFD1, TERA, p97
UniGene
Hs.529782
Gene Chromosome Location
Chr.9: 35056068-35072742 on Build GRCh38
Transcripts
NM_007126.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4752637 c.2098C>T p.R700C Chr.9: 35059126-35059126 on Build GRCh38
1636173 c.2147A>G p.N716S Chr.9: 35059077-35059077 on Build GRCh38
6183263 c.2144C>T p.T715I Chr.9: 35059080-35059080 on Build GRCh38
1108557 c.2110G>A p.E704K Chr.9: 35059114-35059114 on Build GRCh38
5876181 c.2078G>A p.R693H Chr.9: 35059146-35059146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764519589 Chr.9: 35059187-35059187 on Build GRCh38
rs767762212 Chr.9: 35059165-35059165 on Build GRCh38
rs753011642 Chr.9: 35059172-35059172 on Build GRCh38
rs767850418 Chr.9: 35059042-35059042 on Build GRCh38
rs759649712 Chr.9: 35059160-35059160 on Build GRCh38
rs955893615 Chr.9: 35059202-35059202 on Build GRCh38
rs988652510 Chr.9: 35059225-35059225 on Build GRCh38
rs543689812 Chr.9: 35059115-35059115 on Build GRCh38
rs746559433 Chr.9: 35059246-35059246 on Build GRCh38
rs900221571 Chr.9: 35059205-35059205 on Build GRCh38
rs746617777 Chr.9: 35059098-35059098 on Build GRCh38
rs756573088 Chr.9: 35059053-35059053 on Build GRCh38
rs577805218 Chr.9: 35059073-35059073 on Build GRCh38
rs947139359 Chr.9: 35059233-35059233 on Build GRCh38
rs774541798 Chr.9: 35059157-35059157 on Build GRCh38
rs757829124 Chr.9: 35059210-35059210 on Build GRCh38
rs1054911550 Chr.9: 35059046-35059046 on Build GRCh38
rs368358470 Chr.9: 35059051-35059051 on Build GRCh38
rs377125404 Chr.9: 35059076-35059076 on Build GRCh38
rs768462341 Chr.9: 35059104-35059104 on Build GRCh38
rs902184848 Chr.9: 35059097-35059097 on Build GRCh38
rs1000496477 Chr.9: 35059103-35059103 on Build GRCh38
rs541161657 Chr.9: 35059034-35059034 on Build GRCh38
rs769793706 Chr.9: 35059132-35059132 on Build GRCh38
rs754299259 Chr.9: 35059191-35059191 on Build GRCh38
rs878957947 Chr.9: 35059222-35059222 on Build GRCh38
rs372583061 Chr.9: 35059067-35059067 on Build GRCh38
rs1048740862 Chr.9: 35059129-35059130 on Build GRCh38
rs878882315 Chr.9: 35059223-35059223 on Build GRCh38
rs563393296 Chr.9: 35059154-35059154 on Build GRCh38
rs564249854 Chr.9: 35059056-35059056 on Build GRCh38
rs773283632 Chr.9: 35059139-35059139 on Build GRCh38
rs779399540 Chr.9: 35059244-35059244 on Build GRCh38
rs747975110 Chr.9: 35059129-35059129 on Build GRCh38
rs763011046 Chr.9: 35059142-35059142 on Build GRCh38
rs779670266 Chr.9: 35059092-35059092 on Build GRCh38
rs756485118 Chr.9: 35059178-35059178 on Build GRCh38
rs914286524 Chr.9: 35059226-35059226 on Build GRCh38

View Full Product Details