Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3057494-3057719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AES
Entrez Gene ID
166
Gene Name
amino-terminal enhancer of split
Gene Aliases
AES-1, AES-2, ESP1, GRG, GRG5, Grg-5, TLE5
UniGene
Hs.515053
Gene Chromosome Location
Chr.19: 3052910-3063107 on Build GRCh38
Transcripts
NM_001130.5 NM_198969.1 NM_198970.1 XM_006722664.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5056494 c.376C>T p.R126C Chr.19: 3057693-3057693 on Build GRCh38
6316061 c.376C>A p.R126S Chr.19: 3057693-3057693 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370175011 Chr.19: 3057674-3057674 on Build GRCh38
rs376750954 Chr.19: 3057641-3057641 on Build GRCh38
rs536377423 Chr.19: 3057616-3057616 on Build GRCh38
rs538870268 Chr.19: 3057608-3057608 on Build GRCh38
rs777913658 Chr.19: 3057550-3057550 on Build GRCh38
rs571802028 Chr.19: 3057566-3057566 on Build GRCh38
rs747113305 Chr.19: 3057685-3057685 on Build GRCh38
rs750192743 Chr.19: 3057671-3057671 on Build GRCh38
rs758656482 Chr.19: 3057673-3057673 on Build GRCh38
rs983324400 Chr.19: 3057539-3057539 on Build GRCh38
rs975477468 Chr.19: 3057555-3057555 on Build GRCh38
rs1024020101 Chr.19: 3057638-3057638 on Build GRCh38
rs974421547 Chr.19: 3057587-3057587 on Build GRCh38
rs945871185 Chr.19: 3057606-3057606 on Build GRCh38
rs763956103 Chr.19: 3057651-3057651 on Build GRCh38
rs752936219 Chr.19: 3057648-3057648 on Build GRCh38
rs879072795 Chr.19: 3057664-3057664 on Build GRCh38
rs560140295 Chr.19: 3057557-3057557 on Build GRCh38
rs552172887 Chr.19: 3057633-3057634 on Build GRCh38
rs369942753 Chr.19: 3057615-3057615 on Build GRCh38
rs757140015 Chr.19: 3057665-3057665 on Build GRCh38
rs964829848 Chr.19: 3057586-3057586 on Build GRCh38
rs934092877 Chr.19: 3057561-3057561 on Build GRCh38
rs920276008 Chr.19: 3057598-3057598 on Build GRCh38
rs752286739 Chr.19: 3057647-3057647 on Build GRCh38
rs955490803 Chr.19: 3057562-3057562 on Build GRCh38
rs372897754 Chr.19: 3057642-3057642 on Build GRCh38
rs963247683 Chr.19: 3057552-3057552 on Build GRCh38
rs759331874 Chr.19: 3057635-3057642 on Build GRCh38
rs554765471 Chr.19: 3057623-3057623 on Build GRCh38
rs113900847 Chr.19: 3057637-3057637 on Build GRCh38
rs951565355 Chr.19: 3057536-3057536 on Build GRCh38
rs527279876 Chr.19: 3057558-3057558 on Build GRCh38
rs757804185 Chr.19: 3057585-3057585 on Build GRCh38
rs148382760 Chr.19: 3057565-3057565 on Build GRCh38
rs375824445 Chr.19: 3057649-3057649 on Build GRCh38
rs577172054 Chr.19: 3057666-3057666 on Build GRCh38
rs769070431 Chr.19: 3057630-3057630 on Build GRCh38
rs767247887 Chr.19: 3057646-3057646 on Build GRCh38
rs749266780 Chr.19: 3057583-3057583 on Build GRCh38
rs780153150 Chr.19: 3057629-3057629 on Build GRCh38
rs201406195 Chr.19: 3057657-3057657 on Build GRCh38
rs374888556 Chr.19: 3057609-3057609 on Build GRCh38
rs112627745 Chr.19: 3057678-3057678 on Build GRCh38
rs986968692 Chr.19: 3057570-3057570 on Build GRCh38

View Full Product Details