Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 25268746-25268991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK5
Entrez Gene ID
80005
Gene Name
dedicator of cytokinesis 5
Gene Aliases
-
UniGene
Hs.195403
Gene Chromosome Location
Chr.8: 25184723-25415716 on Build GRCh38
Transcripts
NM_001322810.1 NM_024940.7 XM_011544661.1 XM_011544662.1 XM_011544663.2 XM_017013865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7317356 c.137G>C p.R46T Chr.8: 25268854-25268854 on Build GRCh38
4968217 c.168G>C p.K56N Chr.8: 25268885-25268885 on Build GRCh38
5803156 c.149T>A p.L50H Chr.8: 25268866-25268866 on Build GRCh38
750018 c.136A>G p.R46G Chr.8: 25268853-25268853 on Build GRCh38
1098420 c.132G>A p.W44* Chr.8: 25268849-25268849 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770610949 Chr.8: 25268849-25268849 on Build GRCh38
rs77483352 Chr.8: 25268794-25268794 on Build GRCh38
rs767030276 Chr.8: 25268867-25268867 on Build GRCh38
rs1035056974 Chr.8: 25268955-25268955 on Build GRCh38
rs779488861 Chr.8: 25268909-25268909 on Build GRCh38
rs912815855 Chr.8: 25268863-25268863 on Build GRCh38
rs759141209 Chr.8: 25268852-25268852 on Build GRCh38
rs764083786 Chr.8: 25268926-25268926 on Build GRCh38
rs753628706 Chr.8: 25268927-25268927 on Build GRCh38
rs79118805 Chr.8: 25268793-25268793 on Build GRCh38
rs555051315 Chr.8: 25268966-25268966 on Build GRCh38
rs111279151 Chr.8: 25268846-25268846 on Build GRCh38
rs780223269 Chr.8: 25268810-25268810 on Build GRCh38
rs537102701 Chr.8: 25268958-25268958 on Build GRCh38
rs551592508 Chr.8: 25268815-25268815 on Build GRCh38
rs966971768 Chr.8: 25268918-25268918 on Build GRCh38
rs986730924 Chr.8: 25268850-25268850 on Build GRCh38
rs749377441 Chr.8: 25268925-25268925 on Build GRCh38
rs1010994828 Chr.8: 25268858-25268858 on Build GRCh38
rs755599187 Chr.8: 25268895-25268897 on Build GRCh38
rs774024490 Chr.8: 25268851-25268851 on Build GRCh38
rs778039652 Chr.8: 25268825-25268825 on Build GRCh38
rs368499935 Chr.8: 25268785-25268785 on Build GRCh38
rs752187477 Chr.8: 25268869-25268869 on Build GRCh38
rs866374107 Chr.8: 25268878-25268878 on Build GRCh38
rs974586876 Chr.8: 25268783-25268783 on Build GRCh38
rs747473875 Chr.8: 25268814-25268814 on Build GRCh38

View Full Product Details