Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13764448-13764673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRI1
Entrez Gene ID
84245
Gene Name
methylthioribose-1-phosphate isomerase 1
Gene Aliases
MRDI, MTNA, Ypr118w
UniGene
Hs.439370
Gene Chromosome Location
Chr.19: 13764492-13774282 on Build GRCh38
Transcripts
NM_001031727.2 NM_032285.2 XM_011528355.2 XM_011528356.2 XM_011528357.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
709836 c.54G>T p.Q18H Chr.19: 13764642-13764642 on Build GRCh38
4703170 c.14C>T p.A5V Chr.19: 13764602-13764602 on Build GRCh38
7108902 c.18C>T p.I6I Chr.19: 13764606-13764606 on Build GRCh38
4845635 c.35C>A p.S12Y Chr.19: 13764623-13764623 on Build GRCh38
3529220 c.67A>T p.K23* Chr.19: 13764655-13764655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964359949 Chr.19: 13764505-13764505 on Build GRCh38
rs199664561 Chr.19: 13764585-13764585 on Build GRCh38
rs772283715 Chr.19: 13764544-13764544 on Build GRCh38
rs539772625 Chr.19: 13764520-13764520 on Build GRCh38
rs370106395 Chr.19: 13764603-13764603 on Build GRCh38
rs372185077 Chr.19: 13764620-13764620 on Build GRCh38
rs544406642 Chr.19: 13764560-13764560 on Build GRCh38
rs980171832 Chr.19: 13764490-13764491 on Build GRCh38
rs141796284 Chr.19: 13764639-13764639 on Build GRCh38
rs200149723 Chr.19: 13764552-13764552 on Build GRCh38
rs908156125 Chr.19: 13764515-13764515 on Build GRCh38
rs766126995 Chr.19: 13764560-13764561 on Build GRCh38
rs137877795 Chr.19: 13764623-13764623 on Build GRCh38
rs772947643 Chr.19: 13764616-13764616 on Build GRCh38
rs754962859 Chr.19: 13764553-13764553 on Build GRCh38
rs375984822 Chr.19: 13764533-13764533 on Build GRCh38
rs150577856 Chr.19: 13764648-13764648 on Build GRCh38
rs748215741 Chr.19: 13764555-13764555 on Build GRCh38
rs751682443 Chr.19: 13764637-13764637 on Build GRCh38
rs200918680 Chr.19: 13764567-13764567 on Build GRCh38
rs963215189 Chr.19: 13764656-13764656 on Build GRCh38
rs573194689 Chr.19: 13764538-13764538 on Build GRCh38
rs1014246581 Chr.19: 13764479-13764479 on Build GRCh38
rs983708394 Chr.19: 13764491-13764491 on Build GRCh38
rs1011154118 Chr.19: 13764486-13764486 on Build GRCh38
rs765134239 Chr.19: 13764589-13764589 on Build GRCh38
rs983204709 Chr.19: 13764647-13764647 on Build GRCh38
rs753681245 Chr.19: 13764580-13764580 on Build GRCh38
rs767191030 Chr.19: 13764575-13764575 on Build GRCh38
rs562232569 Chr.19: 13764557-13764557 on Build GRCh38
rs777916267 Chr.19: 13764590-13764590 on Build GRCh38
rs575514843 Chr.19: 13764490-13764490 on Build GRCh38
rs760434055 Chr.19: 13764619-13764619 on Build GRCh38
rs747254378 Chr.19: 13764592-13764592 on Build GRCh38
rs781453540 Chr.19: 13764642-13764642 on Build GRCh38
rs760484995 Chr.19: 13764583-13764583 on Build GRCh38
rs773593157 Chr.19: 13764545-13764545 on Build GRCh38
rs368981227 Chr.19: 13764634-13764634 on Build GRCh38
rs891666903 Chr.19: 13764564-13764564 on Build GRCh38
rs375963533 Chr.19: 13764630-13764630 on Build GRCh38
rs557197239 Chr.19: 13764484-13764484 on Build GRCh38
rs932365634 Chr.19: 13764525-13764525 on Build GRCh38
rs762391712 Chr.19: 13764548-13764548 on Build GRCh38
rs770247762 Chr.19: 13764566-13764566 on Build GRCh38
rs767127949 Chr.19: 13764550-13764550 on Build GRCh38
rs79169978 Chr.19: 13764607-13764607 on Build GRCh38
rs746270000 Chr.19: 13764598-13764598 on Build GRCh38
rs983653920 Chr.19: 13764532-13764532 on Build GRCh38
rs900101917 Chr.19: 13764614-13764614 on Build GRCh38
rs562164913 Chr.19: 13764546-13764546 on Build GRCh38
rs750227594 Chr.19: 13764551-13764551 on Build GRCh38
rs184530887 Chr.19: 13764478-13764478 on Build GRCh38
rs200080715 Chr.19: 13764617-13764617 on Build GRCh38
rs751137137 Chr.19: 13764549-13764549 on Build GRCh38
rs974417867 Chr.19: 13764518-13764518 on Build GRCh38
rs765156999 Chr.19: 13764627-13764627 on Build GRCh38
rs777074396 Chr.19: 13764563-13764563 on Build GRCh38
rs777926780 Chr.19: 13764559-13764559 on Build GRCh38
rs952337205 Chr.19: 13764491-13764492 on Build GRCh38
rs781508871 Chr.19: 13764597-13764597 on Build GRCh38
rs1011519325 Chr.19: 13764595-13764595 on Build GRCh38
rs146938415 Chr.19: 13764608-13764608 on Build GRCh38
rs761290425 Chr.19: 13764569-13764569 on Build GRCh38
rs779080974 Chr.19: 13764554-13764554 on Build GRCh38
rs759306075 Chr.19: 13764625-13764625 on Build GRCh38
rs748423476 Chr.19: 13764543-13764543 on Build GRCh38
rs558128979 Chr.19: 13764530-13764530 on Build GRCh38
rs1018560929 Chr.19: 13764495-13764495 on Build GRCh38
rs753785948 Chr.19: 13764586-13764586 on Build GRCh38
rs760070994 Chr.19: 13764565-13764565 on Build GRCh38
rs757578195 Chr.19: 13764594-13764594 on Build GRCh38
rs922274666 Chr.19: 13764523-13764523 on Build GRCh38
rs540678871 Chr.19: 13764540-13764540 on Build GRCh38
rs138769139 Chr.19: 13764588-13764588 on Build GRCh38
rs377020586 Chr.19: 13764580-13764580 on Build GRCh38
rs771299711 Chr.19: 13764562-13764562 on Build GRCh38

View Full Product Details